Product References
A low-carbohydrate diet induces hepatic insulin resistance and metabolic associated fatty liver disease in mice.
Molecular metabolism
Long F,Bhatti MR,Kellenberger A,Sun W,Modica S,Höring M,Liebisch G,Krieger JP,Wolfrum C,Challa TD
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Neutralization
Wed Mar 01 00:00:00 EST 2023
LILRB2/PirB mediates macrophage recruitment in fibrogenesis of nonalcoholic steatohepatitis.
Nature communications
Li DP,Huang L,Kan RR,Meng XY,Wang SY,Zou HJ,Guo YM,Luo PQ,Pan LM,Xiang YX,Mao BB,Xie YY,Wang ZH,Yang M,He R,Yang Y,Liu ZL,Xie JH,Ma DL,Zhang BP,Shao SY,Chen X,Xu SM,He WT,Li WJ,Chen Y,Yu XF
16-7061-81 was used in Neutralization experiments to demonstrate that PirB/LILRB2 regulates the migration of macrophages during NASH by binding with its ligand angiopoietin-like protein 8.
Sat Jul 22 00:00:00 EDT 2023
Lin28B-high breast cancer cells promote immune suppression in the lung pre-metastatic niche via exosomes and support cancer progression.
Nature communications
Qi M,Xia Y,Wu Y,Zhang Z,Wang X,Lu L,Dai C,Song Y,Xu K,Ji W,Zhan L
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Flow Cytometry
Wed Feb 16 00:00:00 EST 2022
Sox2 overexpression alleviates noise-induced hearing loss by inhibiting inflammation-related hair cell apoptosis.
Journal of neuroinflammation
Chen D,Jia G,Zhang Y,Mao H,Zhao L,Li W,Chen Y,Ni Y
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Immunocytochemistry
Mon Feb 28 00:00:00 EST 2022
Sox2 overexpression alleviates noise-induced hearing loss by inhibiting inflammation-related hair cell apoptosis.
Journal of neuroinflammation
Chen D,Jia G,Zhang Y,Mao H,Zhao L,Li W,Chen Y,Ni Y
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Immunocytochemistry
Mon Feb 28 00:00:00 EST 2022
Sox2 overexpression alleviates noise-induced hearing loss by inhibiting inflammation-related hair cell apoptosis.
Journal of neuroinflammation
Chen D,Jia G,Zhang Y,Mao H,Zhao L,Li W,Chen Y,Ni Y
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Immunocytochemistry
Mon Feb 28 00:00:00 EST 2022
Senescent hepatic stellate cells promote liver regeneration through IL-6 and ligands of CXCR2.
JCI insight
Cheng N,Kim KH,Lau LF
16-7061-81 was used in Immunocytochemistry to conclude that HSC senescence, largely induced by CCN1, is a programmed response to PHx and plays a critical role in liver regeneration through signaling pathways activated by IL-6 and ligands of CXCR2.
Thu Jun 16 00:00:00 EDT 2022
Tumor-derived IL-6 and trans-signaling among tumor, fat, and muscle mediate pancreatic cancer cachexia.
The Journal of experimental medicine
Rupert JE,Narasimhan A,Jengelley DHA,Jiang Y,Liu J,Au E,Silverman LM,Sandusky G,Bonetto A,Cao S,Lu X,O'Connell TM,Liu Y,Koniaris LG,Zimmers TA
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Neutralization
Mon Jun 07 00:00:00 EDT 2021
Restoration of 5-methoxytryptophan protects against atherosclerotic chondrogenesis and calcification in ApoE-/- mice fed high fat diet.
Journal of biomedical science
Lee GL,Liao TL,Wu JY,Wu KK,Kuo CC
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in ELISA
Mon Nov 08 00:00:00 EST 2021
TNF-Like Ligand 1 Aberrance Aggravates Nonalcoholic Steatohepatitis via M1 Macrophage Polarization.
Oxidative medicine and cellular longevity
Luo Y,Guo J,Jia W,Wu M,Yin F,Niu G,Shih DQ,Targan SR,Zhang X
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Flow Cytometry
Fri Mar 04 00:00:00 EST 2022
Murine Model of Sinusitis Infection for Screening Antimicrobial and Immunomodulatory Therapies.
Frontiers in cellular and infection microbiology
Alford MA,Choi KG,Trimble MJ,Masoudi H,Kalsi P,Pletzer D,Hancock REW
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in ELISA
Fri Jul 02 00:00:00 EDT 2021
Betulinic Acid Ameliorates the Severity of Acute Pancreatitis via Inhibition of the NF-κB Signaling Pathway in Mice.
International journal of molecular sciences
Zhou Z,Choi JW,Shin JY,Kim DU,Kweon B,Oh H,Kim YC,Song HJ,Bae GS,Park SJ
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Immunocytochemistry
Sat Jun 26 00:00:00 EDT 2021
Spatial proteomics revealed a CX3CL1-dependent crosstalk between the urothelium and relocated macrophages through IL-6 during an acute bacterial infection in the urinary bladder.
Mucosal immunology
Bottek J,Soun C,Lill JK,Dixit A,Thiebes S,Beerlage AL,Horstmann M,Urbanek A,Heuer H,Uszkoreit J,Eisenacher M,Bracht T,Sitek B,Hoffmann F,Vijitha N,von Eggeling F,Engel DR
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Immunohistochemistry
Wed Jul 01 00:00:00 EDT 2020
Spatial proteomics revealed a CX3CL1-dependent crosstalk between the urothelium and relocated macrophages through IL-6 during an acute bacterial infection in the urinary bladder.
Mucosal immunology
Bottek J,Soun C,Lill JK,Dixit A,Thiebes S,Beerlage AL,Horstmann M,Urbanek A,Heuer H,Uszkoreit J,Eisenacher M,Bracht T,Sitek B,Hoffmann F,Vijitha N,von Eggeling F,Engel DR
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Immunohistochemistry
Wed Jul 01 00:00:00 EDT 2020
Spatial proteomics revealed a CX3CL1-dependent crosstalk between the urothelium and relocated macrophages through IL-6 during an acute bacterial infection in the urinary bladder.
Mucosal immunology
Bottek J,Soun C,Lill JK,Dixit A,Thiebes S,Beerlage AL,Horstmann M,Urbanek A,Heuer H,Uszkoreit J,Eisenacher M,Bracht T,Sitek B,Hoffmann F,Vijitha N,von Eggeling F,Engel DR
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Immunohistochemistry
Wed Jul 01 00:00:00 EDT 2020
Vanadium Derivative Exposure Promotes Functional Alterations of VSMCs and Consequent Atherosclerosis via ROS/p38/NF-κB-Mediated IL-6 Production.
International journal of molecular sciences
Yeh CC,Wu JY,Lee GL,Wen HT,Lin P,Kuo CC
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in ELISA
Wed Dec 04 00:00:00 EST 2019
Hepatic inflammation caused by dysregulated bile acid synthesis is reversible by butyrate supplementation.
The Journal of pathology
Sheng L,Jena PK,Hu Y,Liu HX,Nagar N,Kalanetra KM,French SW,French SW,Mills DA,Wan YY
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Western Blot
Fri Dec 01 00:00:00 EST 2017
Depletion of microglia exacerbates postischemic inflammation and brain injury.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Jin WN,Shi SX,Li Z,Li M,Wood K,Gonzales RJ,Liu Q
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
TNFR-Associated Factors 2 and 5 Differentially Regulate the Instructive IL-6 Receptor Signaling Required for Th17 Development.
Journal of immunology (Baltimore, Md. : 1950)
Nagashima H,Okuyama Y,Hayashi T,Ishii N,So T
16-7061-81 was used in Functional assay to demonstrate that both TNFR-associated factor (TRAF)2 and TRAF5 work as important regulators of the IL-6R signalling needed for IL-17-producing CD4(+) T cell development.
Sun May 15 00:00:00 EDT 2016
Beneficial Effects of Anti-Interleukin-6 Antibodies on Impaired Gastrointestinal Motility, Inflammation and Increased Colonic Permeability in a Murine Model of Sepsis Are Most Pronounced When Administered in a Preventive Setup.
PloS one
Nullens S,Staessens M,Peleman C,Plaeke P,Malhotra-Kumar S,Francque S,De Man JG,De Winter BY
16-7061 was used in Functional assays to study the effect of therapeutic or curative administration of anti-IL6 antibodies on overall gastrointestinal motility, colonic permeability and translocation of intestinal bacteria.
Fri Aug 19 00:00:00 EDT 2016
Sprouty1 induces a senescence-associated secretory phenotype by regulating NFκB activity: implications for tumorigenesis.
Cell death and differentiation
Macià A,Vaquero M,Gou-Fàbregas M,Castelblanco E,Valdivielso JM,Anerillas C,Mauricio D,Matias-Guiu X,Ribera J,Encinas M
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Neutralization
Sat Feb 01 00:00:00 EST 2014
Distinct dictation of Japanese encephalitis virus-induced neuroinflammation and lethality via triggering TLR3 and TLR4 signal pathways.
PLoS pathogens
Han YW,Choi JY,Uyangaa E,Kim SB,Kim JH,Kim BS,Kim K,Eo SK
16-7061 was used in an ELISA assay to suggest that a balanced triggering of TLR signal array by viral components during Japanese encephalitis progression could be responsible for determining disease outcome.
Mon Sep 01 00:00:00 EDT 2014
Reduced B lymphoid kinase (Blk) expression enhances proinflammatory cytokine production and induces nephrosis in C57BL/6-lpr/lpr mice.
PloS one
Samuelson EM,Laird RM,Papillion AM,Tatum AH,Princiotta MF,Hayes SM
16-7061 was used in Flow cytometry/Cell sorting to demonstrate how BLK risk alleles confer susceptibility to systemic lupus erythematosus.
Tue Nov 11 00:00:00 EST 2014
Persistently active microbial molecules prolong innate immune tolerance in vivo.
PLoS pathogens
Lu M,Varley AW,Munford RS
16-7061 was used in Flow cytometry/Cell sorting to demonstrate a novel mechanism for tolerance reprogramming following exposure to Gram-negative bacteria.
Wed May 01 00:00:00 EDT 2013
β(2) integrins inhibit TLR responses by regulating NF-κB pathway and p38 MAPK activation.
European journal of immunology
Yee NK,Hamerman JA
16-7061 was used in Flow cytometry/Cell sorting to show that deficiency in β(2) integrins causes hyperresponsiveness to TLR stimulation.
Fri Mar 01 00:00:00 EST 2013
Adhesion to substrates induces dendritic cell endothelization and decreases immunological response.
Immunobiology
Osterbur J,Sprague L,Muccioli M,Pate M,Mansfield K,McGinty J,Li Y,Li Y,Shirure V,Courreges MC,Benencia F
16-7061 was used in an ELISA assay to investigate the effects of different soluble factors, substrates and culture surfaces on the phenotype and function of dendritic cells.
Tue Jan 01 00:00:00 EST 2013
The liver X receptor agonist GW3965 improves recovery from mild repetitive traumatic brain injury in mice partly through apolipoprotein E.
PloS one
Namjoshi DR,Martin G,Donkin J,Wilkinson A,Stukas S,Fan J,Carr M,Tabarestani S,Wuerth K,Hancock RE,Wellington CL
16-7061 was used in an ELISA assay to investigate the therapeutic use of Liver X receptor for treatment of TBI.
Tue Jul 09 00:00:00 EDT 2013
Schistosoma japonicum soluble egg antigens attenuate IFN-γ-induced MHC class II expression in RAW 264.7 macrophages.
PloS one
Tang GX,Zhou HJ,Xu JW,Xu JM,Ji MJ,Wu HW,Wu GL
16-7061 was used in Blocking experiments to elucidate the innate immune-related molecular events involved in the interaction between S. japonicum and its host.
Tue May 07 00:00:00 EDT 2013
Schistosoma japonicum soluble egg antigens attenuate IFN-γ-induced MHC class II expression in RAW 264.7 macrophages.
PloS one
Tang GX,Zhou HJ,Xu JW,Xu JM,Ji MJ,Wu HW,Wu GL
16-7061 was used in Blocking experiments to elucidate the innate immune-related molecular events involved in the interaction between S. japonicum and its host.
Tue May 07 00:00:00 EDT 2013
Signaling by the phosphatase MKP-1 in dendritic cells imprints distinct effector and regulatory T cell fates.
Immunity
Huang G,Wang Y,Shi LZ,Kanneganti TD,Chi H
16-7061 was used in Flow cytometry/Cell sorting to identify a regulatory circuit linking MKP-1 signaling in DCs, production of polarizing cytokines, and integration of DC-derived signals in responding T cells.
Fri Jul 22 00:00:00 EDT 2011
Lysine methylation of the NF-κB subunit RelA by SETD6 couples activity of the histone methyltransferase GLP at chromatin to tonic repression of NF-κB signaling.
Nature immunology
Levy D,Kuo AJ,Chang Y,Schaefer U,Kitson C,Cheung P,Espejo A,Zee BM,Liu CL,Tangsombatvisit S,Tennen RI,Kuo AY,Tanjing S,Cheung R,Chua KF,Utz PJ,Shi X,Prinjha RK,Lee K,Garcia BA,Bedford MT,Tarakhovsky A,Cheng X,Gozani O
16-7061 was used in an ELISA assay to screen candidate protein lysine methyltransferases and identify SETD6 as a methyltransferase that monomethylated chromatin-associated transcription factor NF-κB subunit RelA.
Sat Jan 01 00:00:00 EST 2011
Distinct inflammatory signals have physiologically divergent effects on epigenetic regulation of Foxp3 expression and Treg function.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons
Lal G,Yin N,Xu J,Lin M,Schroppel S,Ding Y,Marie I,Levy DE,Bromberg JS
16-7061 was used in Functional assays to investigate how different inflammatory signals affect Foxp3 expression and Tregs plasticity, showing that distinct signals have physiologically divergent effects on Foxp3 epigenetic regulation.
Tue Feb 01 00:00:00 EST 2011
Synthetic cationic peptide IDR-1002 provides protection against bacterial infections through chemokine induction and enhanced leukocyte recruitment.
Journal of immunology (Baltimore, Md. : 1950)
Nijnik A,Madera L,Ma S,Waldbrook M,Elliott MR,Easton DM,Mayer ML,Mullaly SC,Kindrachuk J,Jenssen H,Hancock RE
16-7061 was used in an ELISA assay to study the importance of the chemokine induction activity of host defence peptides in protection against bacterial infections.
Mon Mar 01 00:00:00 EST 2010
Contrasting roles for all-trans retinoic acid in TGF-beta-mediated induction of Foxp3 and Il10 genes in developing regulatory T cells.
The Journal of experimental medicine
Maynard CL,Hatton RD,Helms WS,Oliver JR,Stephensen CB,Weaver CT
16-7061 was used in Flow cytometry/Cell sorting to investigate alternative roles for all-trans retinoic acid in developing regulatory T cells.
Mon Feb 16 00:00:00 EST 2009
IL-6 produced by immune complex-activated follicular dendritic cells promotes germinal center reactions, IgG responses and somatic hypermutation.
International immunology
Wu Y,El Shikh ME,El Sayed RM,Best AM,Szakal AK,Tew JG
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Neutralization
Mon Jun 01 00:00:00 EDT 2009
IL-6 produced by immune complex-activated follicular dendritic cells promotes germinal center reactions, IgG responses and somatic hypermutation.
International immunology
Wu Y,El Shikh ME,El Sayed RM,Best AM,Szakal AK,Tew JG
Published figure using IL-6 monoclonal antibody (Product # 16-7061-81) in Neutralization
Mon Jun 01 00:00:00 EDT 2009
Maladaptive role of IL-6 in ischemic acute renal failure.
Journal of the American Society of Nephrology : JASN
Kielar ML,John R,Bennett M,Richardson JA,Shelton JM,Chen L,Jeyarajah DR,Zhou XJ,Zhou H,Chiquett B,Nagami GT,Lu CY
16-7061 was used in Functional assays to investigate the role of IL-6 in murine ischemic acute renal failure.
Tue Nov 01 00:00:00 EST 2005