Product References
E-cigarettes and health risks: more to the flavor than just the name.
American journal of physiology. Lung cellular and molecular physiology
Ween MP,Moshensky A,Thredgold L,Bastian NA,Hamon R,Badiei A,Nguyen PT,Herewane K,Jersmann H,Bojanowski CM,Shin J,Reynolds PN,Crotty Alexander LE,Hodge SJ
17-9917 was used in Flow cytometry/Cell sorting to investigate the flavouring profile of 10 different flavoured E-liquids. We assessed bronchial epithelial cell viability and apoptosis,.
Thu Apr 01 00:00:00 EDT 2021
Grindelia squarrosa Extract and Grindelic Acid Modulate Pro-inflammatory Functions of Respiratory Epithelium and Human Macrophages.
Frontiers in pharmacology
Gierlikowska B,Filipek A,Gierlikowski W,Kania D,Stefańska J,Demkow U,Kiss AK
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Sat Feb 06 00:00:00 EST 2021
Comprehensive Cell Surface Antigen Analysis Identifies Transferrin Receptor Protein-1 (CD71) as a Negative Selection Marker for Human Neuronal Cells.
Stem cells (Dayton, Ohio)
Menon V,Thomas R,Elgueta C,Horl M,Osborn T,Hallett PJ,Bartos M,Isacson O,Pruszak J
Tue Oct 01 00:00:00 EDT 2019
Bacterial lipopolysaccharide and antimicrobial LL-37 enhance ICAM-1 expression and NF-κB p65 phosphorylation in senescent endothelial cells.
International journal of molecular medicine
Suzuki K,Ohkuma M,Nagaoka I
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2019
n-butanol extract from Folium isatidis inhibits the lipopolysaccharide-induced downregulation of CXCR1 and CXCR2 on human neutrophils.
Molecular medicine reports
Wu B,Wang L,Jiang L,Dong L,Xu F,Lu Y,Jin J,Wang Z,Liang G,Shan X
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
Butyrate upregulates the TLR4 expression and the phosphorylation of MAPKs and NK-κB in colon cancer cell in vitro.
Oncology letters
Xiao T,Wu S,Yan C,Zhao C,Jin H,Yan N,Xu J,Wu Y,Li C,Shao Q,Xia S
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
Bushfire smoke is pro-inflammatory and suppresses macrophage phagocytic function.
Scientific reports
Hamon R,Tran HB,Roscioli E,Ween M,Jersmann H,Hodge S
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Fri Sep 07 00:00:00 EDT 2018
Bushfire smoke is pro-inflammatory and suppresses macrophage phagocytic function.
Scientific reports
Hamon R,Tran HB,Roscioli E,Ween M,Jersmann H,Hodge S
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Fri Sep 07 00:00:00 EDT 2018
Altered Toll-like receptor expression and function in HPV-associated oropharyngeal carcinoma.
Oncotarget
Tobouti PL,Bolt R,Radhakrishnan R,de Sousa SCOM,Hunter KD
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Tue Jan 02 00:00:00 EST 2018
A low dose lipid infusion is sufficient to induce insulin resistance and a pro-inflammatory response in human subjects.
PloS one
Liang H,Lum H,Alvarez A,Garduno-Garcia JJ,Daniel BJ,Musi N
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Fri Jul 27 00:00:00 EDT 2018
S100A9 induces differentiation of acute myeloid leukemia cells through TLR4.
Blood
Laouedj M,Tardif MR,Gil L,Raquil MA,Lachhab A,Pelletier M,Tessier PA,Barabé F
Thu Apr 06 00:00:00 EDT 2017
Phagocytosis and Inflammation: Exploring the effects of the components of E-cigarette vapor on macrophages.
Physiological reports
Ween MP,Whittall JJ,Hamon R,Reynolds PN,Hodge SJ
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Phagocytosis and Inflammation: Exploring the effects of the components of E-cigarette vapor on macrophages.
Physiological reports
Ween MP,Whittall JJ,Hamon R,Reynolds PN,Hodge SJ
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Pro-inflammatory hepatic macrophages generate ROS through NADPH oxidase 2 via endocytosis of monomeric TLR4-MD2 complex.
Nature communications
Kim SY,Jeong JM,Kim SJ,Seo W,Kim MH,Choi WM,Yoo W,Lee JH,Shim YR,Yi HS,Lee YS,Eun HS,Lee BS,Chun K,Kang SJ,Kim SC,Gao B,Kunos G,Kim HM,Jeong WI
17-9917-42 was used in Flow Cytometry to show that palmitate-stimulated CD11b+F4/80low hepatic infiltrating macrophages generate ROS via dynamin-mediated endocytosis of TLR4 and NOX2, independently from MyD88 and TRIF.
Thu Dec 21 00:00:00 EST 2017
Elevated Toll-Like Receptor-Induced CXCL8 Secretion in Human Blood Basophils from Allergic Donors Is Independent of Toll-Like Receptor Expression Levels.
PloS one
Steiner M,Hawranek T,Schneider M,Ferreira F,Horejs-Hoeck J,Harrer A,Himly M
17-9917 was used in Flow cytometry/Cell sorting to demonstrate that decreased TLR expression and elevated CXCL8 secretion can exacerbate allergic symptoms of parasitic infection.
Tue Jul 26 00:00:00 EDT 2016
Hepatitis C Virus Sensing by Human Trophoblasts Induces Innate Immune Responses and Recruitment of Maternal NK Cells: Potential Implications for Limiting Vertical Transmission.
Journal of immunology (Baltimore, Md. : 1950)
Giugliano S,Petroff MG,Warren BD,Jasti S,Linscheid C,Ward A,Kramer A,Dobrinskikh E,Sheiko MA,Gale M,Golden-Mason L,Winn VD,Rosen HR
17-9917 was used in Flow cytometry/Cell sorting to demonstrate that HCV-RNA sensing by human trophoblasts induces recruitment and activation of innate immune cells.
Thu Oct 15 00:00:00 EDT 2015
TLR4 Expression Is Associated with Left Ventricular Dysfunction in Patients Undergoing Coronary Artery Bypass Surgery.
PloS one
Avlas O,Bragg A,Fuks A,Nicholson JD,Farkash A,Porat E,Aravot D,Levy-Drummer RS,Cohen C,Shainberg A,Arad M,Hochhauser E
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Fri Mar 04 00:00:00 EST 2016
HMGB1 induction of clusterin creates a chemoresistant niche in human prostate tumor cells.
Scientific reports
Zhou J,Chen X,Gilvary DL,Tejera MM,Eksioglu EA,Wei S,Djeu JY
17-9917 was used in Flow cytometry/Cell sorting to study seminal crosstalk between dying and residual live tumour cells during treatment with docetaxel that can result in outgrowth of a chemoresistant population.
Thu Oct 15 00:00:00 EDT 2015
Impaired neutrophil extracellular trap formation: a novel defect in the innate immune system of aged individuals.
Aging cell
Hazeldine J,Harris P,Chapple IL,Grant M,Greenwood H,Livesey A,Sapey E,Lord JM
17-9917 was used in Flow cytometry/Cell sorting to identify that impaired neutrophil extracellular trap formation is a novel defect of innate immunity in older adults.
Fri Aug 01 00:00:00 EDT 2014
The COP II adaptor protein TMED7 is required to initiate and mediate the delivery of TLR4 to the plasma membrane.
Science signaling
Liaunardy-Jopeace A,Bryant CE,Gay NJ
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Tue Jul 29 00:00:00 EDT 2014
Negative regulation of Toll-like receptor 4 signaling by IL-10-dependent microRNA-146b.
Proceedings of the National Academy of Sciences of the United States of America
Curtale G,Mirolo M,Renzi TA,Rossato M,Bazzoni F,Locati M
17-9917 was used in Flow cytometry/Cell sorting to identify miR-146b as an IL-10-responsive miR with an anti-inflammatory activity based on multiple targeting of components of the TLR4 pathway in monocytes.
Tue Jul 09 00:00:00 EDT 2013
Body fluid exosomes promote secretion of inflammatory cytokines in monocytic cells via Toll-like receptor signaling.
The Journal of biological chemistry
Bretz NP,Ridinger J,Rupp AK,Rimbach K,Keller S,Rupp C,Marmé F,Umansky L,Umansky V,Eigenbrod T,Sammar M,Altevogt P
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Fri Dec 20 00:00:00 EST 2013
Body fluid exosomes promote secretion of inflammatory cytokines in monocytic cells via Toll-like receptor signaling.
The Journal of biological chemistry
Bretz NP,Ridinger J,Rupp AK,Rimbach K,Keller S,Rupp C,Marmé F,Umansky L,Umansky V,Eigenbrod T,Sammar M,Altevogt P
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Fri Dec 20 00:00:00 EST 2013
Bone marrow mononuclear cells up-regulate toll-like receptor expression and produce inflammatory mediators in response to cigarette smoke extract.
PloS one
Zhou J,Eksioglu EA,Fortenbery NR,Chen X,Wang H,Epling-Burnette PK,Djeu JY,Wei S
17-9917 was used in Flow cytometry/Cell sorting to elucidate the effects of cigarette smoke on human bone marrow haematopoiesis and characterise the inflammatory response.
Thu Dec 01 00:00:00 EST 2011
Candesartan inhibits Toll-like receptor expression and activity both in vitro and in vivo.
Atherosclerosis
Dasu MR,Riosvelasco AC,Jialal I
Published figure using CD284 (TLR4) monoclonal antibody (Product # 17-9917-42) in Flow Cytometry
Thu Jan 01 00:00:00 EST 2009
Human heat shock protein 60 stimulates vascular smooth muscle cell proliferation through Toll-like receptors 2 and 4.
Microbes and infection
de Graaf R,Kloppenburg G,Kitslaar PJ,Bruggeman CA,Stassen F
Thu Jun 01 00:00:00 EDT 2006
Expression and involvement of Toll-like receptors (TLR)2, TLR4, and CD14 in monocyte TNF-alpha production induced by lipopolysaccharides from Neisseria meningitidis.
Medical science monitor : international medical journal of experimental and clinical research
Mirlashari MR,Lyberg T
Fri Aug 01 00:00:00 EDT 2003