Product References
The ribosomal protein S6 kinase alpha-1 (RPS6KA1) induces resistance to venetoclax/azacitidine in acute myeloid leukemia.
Leukemia
Weidenauer K,Schmidt C,Rohde C,Pauli C,Blank MF,Heid D,Waclawiczek A,Corbacioglu A,Göllner S,Lotze M,Vierbaum L,Renders S,Krijgsveld J,Raffel S,Sauer T,Trumpp A,Pabst C,Müller-Tidow C,Janssen M
25-0338-42 was used in Immunocytochemistry to suggest RPS6KA1 as mediator of resistance towards venetoclax/azacitidine and additional RPS6KA1 inhibition as strategy to prevent or overcome resistance.
Tue Aug 01 00:00:00 EDT 2023
Myeloid cell subsets that express latency-associated peptide promote cancer growth by modulating T cells.
iScience
Gabriely G,Ma D,Siddiqui S,Sun L,Skillin NP,Abou-El-Hassan H,Moreira TG,Donnelly D,da Cunha AP,Fujiwara M,Walton LR,Patel A,Krishnan R,Levine SS,Healy BC,Rezende RM,Murugaiyan G,Weiner HL
25-0338-42 was used in Flow Cytometry to identify a previously unknown function by which LAPHi MCs promote tumor growth and offer therapeutic intervention to target these cells in cancer.
Fri Nov 19 00:00:00 EST 2021
Celiac Disease Defined by Over-Sensitivity to Gliadin Activation and Superior Antigen Presentation of Dendritic Cells.
International journal of molecular sciences
Hudec M,Riegerová K,Pala J,Kútna V,Černá M,O Leary VB
Published figure using CD33 monoclonal antibody (Product # 25-0338-42) in Flow Cytometry
Wed Sep 15 00:00:00 EDT 2021
Using antagonistic pleiotropy to design a chemotherapy-induced evolutionary trap to target drug resistance in cancer.
Nature genetics
Lin KH,Rutter JC,Xie A,Pardieu B,Winn ET,Bello RD,Forget A,Itzykson R,Ahn YR,Dai Z,Sobhan RT,Anderson GR,Singleton KR,Decker AE,Winter PS,Locasale JW,Crawford L,Puissant A,Wood KC
25-0338 was used in Flow cytometry/Cell sorting to perform pooled CRISPR-Cas9 knockout screens in acute myeloid leukemia (AML) cells treated with various chemotherapies to map the drug-dependent genetic basis of fitness trade-offs, a concept known as antagonistic pleiotropy (AP).
Wed Apr 01 00:00:00 EDT 2020
Galectin-9 promotes a suppressive microenvironment in human cancer by enhancing STING degradation.
Oncogenesis
Zhang CX,Huang DJ,Baloche V,Zhang L,Xu JX,Li BW,Zhao XR,He J,Mai HQ,Chen QY,Zhang XS,Busson P,Cui J,Li J
Published figure using CD33 monoclonal antibody (Product # 25-0338-42) in Flow Cytometry
Mon Jul 06 00:00:00 EDT 2020
Connecting METTL3 and intratumoural CD33+ MDSCs in predicting clinical outcome in cervical cancer.
Journal of translational medicine
Ni HH,Zhang L,Huang H,Dai SQ,Li J
Published figure using CD33 monoclonal antibody (Product # 25-0338-42) in Flow Cytometry
Thu Oct 15 00:00:00 EDT 2020
iPSC-Derived Platelets Depleted of HLA Class I Are Inert to Anti-HLA Class I and Natural Killer Cell Immunity.
Stem cell reports
Suzuki D,Flahou C,Yoshikawa N,Stirblyte I,Hayashi Y,Sawaguchi A,Akasaka M,Nakamura S,Higashi N,Xu H,Matsumoto T,Fujio K,Manz MG,Hotta A,Takizawa H,Eto K,Sugimoto N
Published figure using CD33 monoclonal antibody (Product # 25-0338-42) in Flow Cytometry
Tue Jan 14 00:00:00 EST 2020
Generation of Human Immunosuppressive Myeloid Cell Populations in Human Interleukin-6 Transgenic NOG Mice.
Frontiers in immunology
Hanazawa A,Ito R,Katano I,Kawai K,Goto M,Suemizu H,Kawakami Y,Ito M,Takahashi T
25-0338 was used in Flow cytometry/Cell sorting to suggest that functional human TAMs can be developed in NOG-hIL-6 Tg mice.
Thu Apr 18 00:00:00 EDT 2019
Exosomal transfer of mitochondria from airway myeloid-derived regulatory cells to T cells.
Redox biology
Hough KP,Trevor JL,Strenkowski JG,Wang Y,Chacko BK,Tousif S,Chanda D,Steele C,Antony VB,Dokland T,Ouyang X,Zhang J,Duncan SR,Thannickal VJ,Darley-Usmar VM,Deshane JS
25-0338 was used in Flow cytometry/Cell sorting to support a potential novel mechanism of cell-cell communication involving exosomal transfer of mitochondria and the bioenergetic and/or redox regulation of target cells.
Sat Sep 01 00:00:00 EDT 2018
The creatine kinase pathway is a metabolic vulnerability in EVI1-positive acute myeloid leukemia.
Nature medicine
Fenouille N,Bassil CF,Ben-Sahra I,Benajiba L,Alexe G,Ramos A,Pikman Y,Conway AS,Burgess MR,Li Q,Luciano F,Auberger P,Galinsky I,DeAngelo DJ,Stone RM,Zhang Y,Perkins AS,Shannon K,Hemann MT,Puissant A,Stegmaier K
25-0338 was used in Flow cytometry/Cell sorting to investigate the effect of MECOM on cellular metabolism.
Wed Mar 01 00:00:00 EST 2017
Surrogate in vitro activation of innate immunity synergizes with interleukin-7 to unleash rapid antigen-driven outgrowth of CD4+ and CD8+ human peripheral blood T-cells naturally recognizing MUC1, HER2/neu and other tumor-associated antigens.
Oncotarget
Pathangey LB,McCurry DB,Gendler SJ,Dominguez AL,Gorman JE,Pathangey G,Mihalik LA,Dang Y,Disis ML,Cohen PA
Published figure using CD33 monoclonal antibody (Product # 25-0338-42) in Flow Cytometry
Tue Feb 14 00:00:00 EST 2017
Antithymocyte Globulin at Clinically Relevant Concentrations Kills Leukemic Blasts.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation
Dabas R,Lee R,Servito MT,Dharmani-Khan P,Modi M,van Slyke T,Luider J,Durand C,Larratt L,Brandwein J,Morris D,Daly A,Khan FM,Storek J
Sun May 01 00:00:00 EDT 2016
Genetic Variability Overrides the Impact of Parental Cell Type and Determines iPSC Differentiation Potential.
Stem cell reports
Kyttälä A,Moraghebi R,Valensisi C,Kettunen J,Andrus C,Pasumarthy KK,Nakanishi M,Nishimura K,Ohtaka M,Weltner J,Van Handel B,Parkkonen O,Sinisalo J,Jalanko A,Hawkins RD,Woods NB,Otonkoski T,Trokovic R
Published figure using CD33 monoclonal antibody (Product # 25-0338-42) in Flow Cytometry
Tue Feb 09 00:00:00 EST 2016
Metabolic regulation of hepatitis B immunopathology by myeloid-derived suppressor cells.
Nature medicine
Pallett LJ,Gill US,Quaglia A,Sinclair LV,Jover-Cobos M,Schurich A,Singh KP,Thomas N,Das A,Chen A,Fusai G,Bertoletti A,Cantrell DA,Kennedy PT,Davies NA,Haniffa M,Maini MK
Published figure using CD33 monoclonal antibody (Product # 25-0338-42) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
Defining human dendritic cell progenitors by multiparametric flow cytometry.
Nature protocols
Breton G,Lee J,Liu K,Nussenzweig MC
Published figure using CD33 monoclonal antibody (Product # 25-0338-42) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Replication of Salmonella enterica Serovar Typhimurium in Human Monocyte-Derived Macrophages.
Infection and immunity
Lathrop SK,Binder KA,Starr T,Cooper KG,Chong A,Carmody AB,Steele-Mortimer O
25-0338 was used in Flow cytometry/Cell sorting to assess the ability of Salmonella to replicate in human macrophages.
Wed Jul 01 00:00:00 EDT 2015
A novel autosomal recessive TERT T1129P mutation in a dyskeratosis congenita family leads to cellular senescence and loss of CD34+ hematopoietic stem cells not reversible by mTOR-inhibition.
Aging
Stockklausner C,Raffel S,Klermund J,Bandapalli OR,Beier F,Brümmendorf TH,Bürger F,Sauer SW,Hoffmann GF,Lorenz H,Tagliaferri L,Nowak D,Hofmann WK,Buergermeister R,Kerber C,Rausch T,Korbel JO,Luke B,Trumpp A,Kulozik AE
25-0338 was used in Flow cytometry/Cell sorting to outline limitations of a potential dyskeratosis congenita therapy based on rapamycin.
Sun Nov 01 00:00:00 EDT 2015
SYK regulates mTOR signaling in AML.
Leukemia
Carnevale J,Ross L,Puissant A,Banerji V,Stone RM,DeAngelo DJ,Ross KN,Stegmaier K
25-0338 was used in Flow cytometry/Cell sorting to study the regulation of mTOR signaling in AML by SYK.
Fri Nov 01 00:00:00 EDT 2013
Targeting CDK1 promotes FLT3-activated acute myeloid leukemia differentiation through C/EBPα.
The Journal of clinical investigation
Radomska HS,Alberich-Jordà M,Will B,Gonzalez D,Delwel R,Tenen DG
Published figure using CD33 monoclonal antibody (Product # 25-0338-42) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2012
Characterization of a novel population of low-density granulocytes associated with disease severity in HIV-1 infection.
PloS one
Cloke T,Munder M,Taylor G,Müller I,Kropf P
25-0338 was used in Flow cytometry/Cell sorting to reveal that increased frequencies of LDGs correlate with disease severity in HIV+ patients.
Wed May 01 00:00:00 EDT 2013