Product References
R274X-mutated Phf6 increased the self-renewal and skewed T cell differentiation of hematopoietic stem cells.
iScience
Lan Y,Yuan S,Guo T,Hou S,Zhao F,Yang W,Cao Y,Chu Y,Jiang E,Yuan W,Wang X
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Fri Jun 16 00:00:00 EDT 2023
LINC01123 promotes immune escape by sponging miR-214-3p to regulate B7-H3 in head and neck squamous-cell carcinoma.
Cell death & disease
Li H,Yang Z,Yang X,Zhang F,Wang J,Wu Z,Wanyan C,Meng Q,Gao W,Yang X,Wei J
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Thu Feb 03 00:00:00 EST 2022
PRDM1/BLIMP1 induces cancer immune evasion by modulating the USP22-SPI1-PD-L1 axis in hepatocellular carcinoma cells.
Nature communications
Li Q,Zhang L,You W,Xu J,Dai J,Hua D,Zhang R,Yao F,Zhou S,Huang W,Dai Y,Zhang Y,Baheti T,Qian X,Pu L,Xu J,Xia Y,Zhang C,Tang J,Wang X
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Mon Dec 12 00:00:00 EST 2022
Baricitinib treatment resolves lower-airway macrophage inflammation and neutrophil recruitment in SARS-CoV-2-infected rhesus macaques.
Cell
Hoang TN,Pino M,Boddapati AK,Viox EG,Starke CE,Upadhyay AA,Gumber S,Nekorchuk M,Busman-Sahay K,Strongin Z,Harper JL,Tharp GK,Pellegrini KL,Kirejczyk S,Zandi K,Tao S,Horton TR,Beagle EN,Mahar EA,Lee MYH,Cohen J,Jean SM,Wood JS,Connor-Stroud F,Stammen RL,Delmas OM,Wang S,Cooney KA,Sayegh MN,Wang L,Filev PD,Weiskopf D,Silvestri G,Waggoner J,Piantadosi A,Kasturi SP,Al-Shakhshir H,Ribeiro SP,Sekaly RP,Levit RD,Estes JD,Vanderford TH,Schinazi RF,Bosinger SE,Paiardini M
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Thu Jan 21 00:00:00 EST 2021
Membrane lymphotoxin-α2β is a novel tumor necrosis factor (TNF) receptor 2 (TNFR2) agonist.
Cell death & disease
Kucka K,Lang I,Zhang T,Siegmund D,Medler J,Wajant H
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Tue Apr 06 00:00:00 EDT 2021
NKD2 mediates stimulation-dependent ORAI1 trafficking to augment Ca2+ entry in T cells.
Cell reports
Wu B,Woo JS,Vila P,Jew M,Leung J,Sun Z,Srikanth S,Gwack Y
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Tue Aug 24 00:00:00 EDT 2021
Development of an Inflammatory CD14+ Dendritic Cell Subset in Humanized Mice.
Frontiers in immunology
Iwabuchi R,Ide K,Terahara K,Wagatsuma R,Iwaki R,Matsunaga H,Tsunetsugu-Yokota Y,Takeyama H,Takahashi Y
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Mon Sep 27 00:00:00 EDT 2021
Rejuvenating Effector/Exhausted CAR T Cells to Stem Cell Memory-Like CAR T Cells By Resting Them in the Presence of CXCL12 and the NOTCH Ligand.
Cancer research communications
Ando M,Kondo T,Tomisato W,Ito M,Shichino S,Srirat T,Mise-Omata S,Nakagawara K,Yoshimura A
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
Inhibition of Cholesterol Esterification Enzyme Enhances the Potency of Human Chimeric Antigen Receptor T Cells against Pancreatic Carcinoma.
Molecular therapy oncolytics
Zhao L,Liu Y,Zhao F,Jin Y,Feng J,Geng R,Sun J,Kang L,Yu L,Wei Y
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Fri Mar 27 00:00:00 EDT 2020
Abrogation of HLA surface expression using CRISPR/Cas9 genome editing: a step toward universal T cell therapy.
Scientific reports
Lee J,Sheen JH,Lim O,Lee Y,Ryu J,Shin D,Kim YY,Kim M
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Tue Oct 20 00:00:00 EDT 2020
Ghrelin ameliorates tumor-induced adipose tissue atrophy and inflammation via Ghrelin receptor-dependent and -independent pathways.
Oncotarget
Liu H,Luo J,Guillory B,Chen JA,Zang P,Yoeli JK,Hernandez Y,Lee II,Anderson B,Storie M,Tewnion A,Garcia JM
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Immunohistochemistry
Tue Sep 01 00:00:00 EDT 2020
Mucosal-associated invariant T (MAIT) cells mediate protective host responses in sepsis.
eLife
Trivedi S,Labuz D,Anderson CP,Araujo CV,Blair A,Middleton EA,Jensen O,Tran A,Mulvey MA,Campbell RA,Hale JS,Rondina MT,Leung DT
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Mon Nov 09 00:00:00 EST 2020
CAIX Regulates GBM Motility and TAM Adhesion and Polarization through EGFR/STAT3 under Hypoxic Conditions.
International journal of molecular sciences
Huang BR,Liu YS,Lai SW,Lin HJ,Shen CK,Yang LY,Lu DY
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Fri Aug 14 00:00:00 EDT 2020
TIGIT and PD-1 Mark Intratumoral T Cells with Reduced Effector Function in B-cell Non-Hodgkin Lymphoma.
Cancer immunology research
Josefsson SE,Beiske K,Blaker YN,Førsund MS,Holte H,Østenstad B,Kimby E,Köksal H,Wälchli S,Bai B,Smeland EB,Levy R,Kolstad A,Huse K,Myklebust JH
53-7349 was used in Flow cytometry/Cell sorting to show that TIGIT and PD-1 mark dysfunctional T cells and suggest that TIGIT and PD-1 coblockade should be further explored to elicit potent antitumor responses in patients with NHL.
Fri Mar 01 00:00:00 EST 2019
Characterization of Antigenic MHC-Class-I-Restricted T Cell Epitopes in the Glycoprotein of Ebolavirus.
Cell reports
Powlson J,Wright D,Zeltina A,Giza M,Nielsen M,Rampling T,Venkatrakaman N,Bowden TA,Hill AVS,Ewer KJ
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Tue Nov 26 00:00:00 EST 2019
Highly efficient multiplex human T cell engineering without double-strand breaks using Cas9 base editors.
Nature communications
Webber BR,Lonetree CL,Kluesner MG,Johnson MJ,Pomeroy EJ,Diers MD,Lahr WS,Draper GM,Slipek NJ,Smeester BA,Lovendahl KN,McElroy AN,Gordon WR,Osborn MJ,Moriarity BS
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Tue Nov 19 00:00:00 EST 2019
Heterogeneous Tumor-Immune Microenvironments among Differentially Growing Metastases in an Ovarian Cancer Patient.
Cell
Jiménez-Sánchez A,Memon D,Pourpe S,Veeraraghavan H,Li Y,Vargas HA,Gill MB,Park KJ,Zivanovic O,Konner J,Ricca J,Zamarin D,Walther T,Aghajanian C,Wolchok JD,Sala E,Merghoub T,Snyder A,Miller ML
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Thu Aug 24 00:00:00 EDT 2017
The distribution and function of human memory T cell subsets in lung cancer.
Immunologic research
Sheng SY,Gu Y,Lu CG,Zou JY,Hong H,Wang R
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Application of a whole blood mycobacterial growth inhibition assay to study immunity against Mycobacterium tuberculosis in a high tuberculosis burden population.
PloS one
Baguma R,Penn-Nicholson A,Smit E,Erasmus M,Day J,Makhethe L,de Kock M,Hughes EJ,van Rooyen M,Pienaar B,Stone L,Hanekom W,Brennan MJ,Wallis RS,Hatherill M,Scriba TJ
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Wed Oct 18 00:00:00 EDT 2017
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Tue Aug 12 00:00:00 EDT 2014
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Tue Aug 12 00:00:00 EDT 2014
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Tue Aug 12 00:00:00 EDT 2014
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Tue Aug 12 00:00:00 EDT 2014
Cell-mediated immune responses to respiratory syncytial virus infection: magnitude, kinetics, and correlates with morbidity and age.
Human vaccines & immunotherapeutics
Geevarghese B,Weinberg A
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Mon Apr 25 00:00:00 EDT 2016
Functional OCT4-specific CD4+ and CD8+ T cells in healthy controls and ovarian cancer patients.
Oncoimmunology
Di J,Massuger LF,Duiveman-de Boer T,Zusterzeel PL,Figdor CG,Torensma R
Published figure using TNF alpha monoclonal antibody (Product # 53-7349-42) in Flow Cytometry
Wed May 01 00:00:00 EDT 2013