Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: BMS180 (JES6-5H4) is raised against recombinant full length IL-2 of mouse origin and recommended for detection of recombinant and native IL-2 of mouse origin by IP,ELISA, FC, WB. The antibody was purified from tissue culture supernatant by protein G column chromatography.
Applications Tested: ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blotting.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-IL-2; il 2; IL-2; ILN; Interleukin; Interleukin-2; Interleukin2; Involved in regulation of T cell clonal expansion; M-IL-2; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Gene Aliases: Il-2; Il2
UniProt ID: (Mouse) P04351
Entrez Gene ID: (Mouse) 16183
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support