Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FAM153A | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_173663.3 | NP_775934.3 | - | - | - |
XM_005265889.3 | XP_005265946.1 | - | - | - | |
XM_006714848.3 | XP_006714911.1 | - | - | - | |
XM_006714849.3 | XP_006714912.1 | - | - | - | |
XM_011534520.2 | XP_011532822.1 | - | - | - | |
XM_011534521.2 | XP_011532823.1 | - | - | - | |
XM_011534522.2 | XP_011532824.1 | - | - | - | |
XM_011534523.2 | XP_011532825.1 | - | - | - | |
XM_011534528.2 | XP_011532830.1 | - | - | - | |
XM_017009352.1 | XP_016864841.1 | - | - | - | |
XM_017009353.1 | XP_016864842.1 | - | - | - | |
XM_017009354.1 | XP_016864843.1 | - | - | - | |
XM_017009355.1 | XP_016864844.1 | - | - | - | |
XM_017009356.1 | XP_016864845.1 | - | - | - | |
XM_017009357.1 | XP_016864846.1 | - | - | - | |
XM_017009358.1 | XP_016864847.1 | - | - | - | |
XM_017009359.1 | XP_016864848.1 | - | - | - | |
XM_017009360.1 | XP_016864849.1 | - | - | - | |
XM_017009361.1 | XP_016864850.1 | - | - | - | |
XM_017009362.1 | XP_016864851.1 | - | - | - | |
XM_017009363.1 | XP_016864852.1 | - | - | - | |
XM_017009364.1 | XP_016864853.1 | - | - | - | |
XM_017009365.1 | XP_016864854.1 | - | - | - | |
XM_017009366.1 | XP_016864855.1 | - | - | - | |
XM_017009367.1 | XP_016864856.1 | - | - | - | |
XM_017009368.1 | XP_016864857.1 | - | - | - | |
XM_017009369.1 | XP_016864858.1 | - | - | - | |
XM_017009370.1 | XP_016864859.1 | - | - | - | |
XM_017009371.1 | XP_016864860.1 | - | - | - | |
GenBank | AB018295.1 | - | - | - | |
AF155097.1 | AAD42863.1 | - | - | - | |
AK289518.1 | - | - | - | ||
DB046513.1 | - | - | - | ||
ENST00000360669.0 | - | - | - |
Literature Links: |
FAM153A PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 400-700 bp |