Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATRX | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_000489.4 | NP_000480.3 | - | - | - |
NM_138270.3 | NP_612114.2 | - | - | - | |
XM_005262153.4 | XP_005262210.2 | - | - | - | |
XM_005262154.4 | XP_005262211.2 | - | - | - | |
XM_005262155.4 | XP_005262212.2 | - | - | - | |
XM_005262156.4 | XP_005262213.2 | - | - | - | |
XM_005262157.4 | XP_005262214.2 | - | - | - | |
XM_006724666.3 | XP_006724729.1 | - | - | - | |
XM_006724667.3 | XP_006724730.1 | - | - | - | |
XM_017029601.1 | XP_016885090.1 | - | - | - | |
XM_017029602.1 | XP_016885091.1 | - | - | - | |
XM_017029603.1 | XP_016885092.1 | - | - | - | |
XM_017029604.1 | XP_016885093.1 | - | - | - | |
XM_017029605.1 | XP_016885094.1 | - | - | - | |
XM_017029606.1 | XP_016885095.1 | - | - | - | |
XM_017029607.1 | XP_016885096.1 | - | - | - | |
XM_017029608.1 | XP_016885097.1 | - | - | - | |
XM_017029609.1 | XP_016885098.1 | - | - | - | |
XM_017029610.1 | XP_016885099.1 | - | - | - | |
XM_017029611.1 | XP_016885100.1 | - | - | - | |
GenBank | AA484729.1 | - | - | - | |
BE927791.1 | - | - | - | ||
BQ189245.1 | - | - | - | ||
ENST00000373344.0 | - | - | - | ||
U72936.2 | AAB49969.1 | - | - | - | |
U72937.2 | AAB49970.2 | - | - | - | |
U72938.2 | AAB49971.2 | - | - | - |
Literature Links: |
ATRX PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 400-700 bp |