Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
DDB2
Entrez Gene ID
1643
Gene Name:
damage specific DNA binding protein 2
Gene Aliases
DDBB, UV-DDB2, XPE
Gene Chromosome Location
Chr.11: on Build GRCh38
UniGene
Hs.700338
Species:
Human
Primer Chromosome Location
Chr.11: 47232775-47232905 on Build GRCh38
View in UCSC browser
Amplicon Length
131
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00516823_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
DDB2
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_000107.2
NP_000098.1
6363556
c.521C>T
p.A174V
299932
c.537A>T
p.G179G
7122595
c.527C>T
p.S176L
27597
c.462_463delAG
p.A155fs*10
6405436
c.511C>A
p.Q171K
1298088
c.501C>T
p.L167L
1298087
c.499C>T
p.L167F
3809304
c.519C>T
p.Y173Y
NM_001300734.1
NP_001287663.1
-
-
-
GenBank
AB107037.1
BAD12557.1
-
-
-
AB107038.1
BAD12558.1
-
-
-
AB107039.1
BAD12559.1
-
-
-
AB107040.1
BAD12560.1
-
-
-
AK091640.1
-
-
-
AK313262.1
-
-
-
BC000093.2
AAH00093.1
-
-
-
BC050455.1
-
-
-
BT007139.1
AAP35803.1
-
-
-
ENST00000256996.0
-
-
-
U18300.1
AAB07897.1
-
-
-
Phenotype
MIM: 600811
Literature Links:
DDB2
PubMed Links
SNP ID
rs1005757127
rs201703288
rs778435738
rs375645261
rs781686709
rs964277772
rs748993672
rs370540741
rs770406867
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.11: 47232837-47232837 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.11: 47232868-47232868 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global
G (0.00)
(1.00)
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS
G (0.00)
(1.00)
African American - Not Available
YRI (Yoruba) - Not Available
SAS
G (0.00)
(1.00)
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR
G (0.00)
(1.00)
Japanese - Not Available
JPT (Japanese) - Not Available
EUR
G (0.00)
(1.00)
AMR
G (0.00)
(1.00)
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.11: 47232865-47232865 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.11: 47232807-47232807 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.11: 47232799-47232799 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.11: 47232877-47232877 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.11: 47232853-47232853 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.11: 47232831-47232831 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.11: 47232859-47232859 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
DNA metabolism protein
damaged DNA-binding protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
nucleotide-excision repair, DNA damage recognition
nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair
pyrimidine dimer repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
response to UV
nucleotide-excision repair, DNA incision
histone H2A monoubiquitination
protein autoubiquitination
global genome nucleotide-excision repair
UV-damage excision repair
DNA binding
damaged DNA binding
ubiquitin-protein transferase activity
protein binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0