Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FGFR2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_000141.4 | NP_000132.3 | 6913198 | c.2329G>A | p.E777K |
2056120 | c.2415C>T | p.Y805Y | |||
1584824 | c.2307C>A | p.Y769* | |||
5608792 | c.2419C>T | p.P807S | |||
5769699 | c.2383G>T | p.D795Y | |||
5370157 | c.2393T>A | p.F798Y | |||
4994841 | c.2371T>A | p.S791T | |||
5812031 | c.2331A>G | p.E777E | |||
6191520 | c.2353G>C | p.D785H | |||
6967027 | c.2401G>T | p.D801Y | |||
3434714 | c.2387C>T | p.S796F | |||
7213305 | c.2392T>A | p.F798I | |||
3434718 | c.2310G>A | p.L770L | |||
5885143 | c.2322A>G | p.Q774Q | |||
5836370 | c.2371delT | p.S791fs*25 | |||
5596722 | c.2411C>T | p.P804L | |||
3866647 | c.2324C>T | p.P775L | |||
7231919 | c.2314C>T | p.L772F | |||
6566451 | c.2400A>T | p.P800P | |||
5608758 | c.2302G>A | p.E768K | |||
6663718 | c.2416G>A | p.E806K | |||
4437260 | c.2323C>T | p.P775S | |||
6006342 | c.2332C>T | p.Q778* | |||
1584826 | c.2343T>C | p.P781P | |||
6848585 | c.2372C>T | p.S791F | |||
5608805 | c.2307C>T | p.Y769Y | |||
1646242 | c.2357C>A | p.T786K | |||
6934053 | c.2383G>A | p.D795N | |||
6128320 | c.2409G>T | p.M803I | |||
6932236 | c.2302G>T | p.E768* | |||
3434716 | c.2311G>A | p.D771N | |||
3434712 | c.2396C>T | p.S799F | |||
5818080 | c.2318G>A | p.S773N | |||
29827 | c.2308T>G | p.L770V | |||
4811851 | c.2339C>T | p.S780L | |||
NM_001144914.1 | NP_001138386.1 | - | - | - | |
NM_001144915.1 | NP_001138387.1 | - | - | - | |
NM_001144916.1 | NP_001138388.1 | - | - | - | |
NM_001144917.1 | NP_001138389.1 | - | - | - | |
NM_001144918.1 | NP_001138390.1 | - | - | - | |
NM_001320654.1 | NP_001307583.1 | - | - | - | |
NM_001320658.1 | NP_001307587.1 | - | - | - | |
NM_022970.3 | NP_075259.4 | 6848584 | c.2375C>T | p.S792F | |
4437259 | c.2326C>T | p.P776S | |||
5608804 | c.2310C>T | p.Y770Y | |||
1146406 | c.2360C>A | p.T787K | |||
7213304 | c.2395T>A | p.F799I | |||
5370156 | c.2396T>A | p.F799Y | |||
5812030 | c.2334A>G | p.E778E | |||
7231918 | c.2317C>T | p.L773F | |||
6913197 | c.2332G>A | p.E778K | |||
5836369 | c.2374delT | p.S792fs*25 | |||
1584825 | c.2346T>C | p.P782P | |||
6566450 | c.2403A>T | p.P801P | |||
6663717 | c.2419G>A | p.E807K | |||
4994840 | c.2374T>A | p.S792T | |||
3434715 | c.2314G>A | p.D772N | |||
3434711 | c.2399C>T | p.S800F | |||
6932235 | c.2305G>T | p.E769* | |||
6128319 | c.2412G>T | p.M804I | |||
5608791 | c.2422C>T | p.P808S | |||
6934052 | c.2386G>A | p.D796N | |||
3866646 | c.2327C>T | p.P776L | |||
2056119 | c.2418C>T | p.Y806Y | |||
6967026 | c.2404G>T | p.D802Y | |||
5769698 | c.2386G>T | p.D796Y | |||
5818079 | c.2321G>A | p.S774N | |||
6973530 | c.2311T>G | p.L771V | |||
5885444 | c.2325A>G | p.Q775Q | |||
5596721 | c.2414C>T | p.P805L | |||
1297002 | c.2342C>T | p.S781L | |||
1584823 | c.2310C>A | p.Y770* | |||
5608757 | c.2305G>A | p.E769K | |||
3434713 | c.2390C>T | p.S797F | |||
1134978 | c.2371T>G | p.C791G | |||
3434717 | c.2313G>A | p.L771L | |||
NM_023029.2 | NP_075418.1 | - | - | - | |
NR_073009.1 | - | - | - | ||
XM_006717708.2 | XP_006717771.1 | - | - | - | |
XM_006717710.3 | XP_006717773.1 | - | - | - | |
XM_006717711.2 | XP_006717774.1 | - | - | - | |
XM_006717712.2 | XP_006717775.1 | - | - | - | |
XM_017015920.1 | XP_016871409.1 | - | - | - | |
XM_017015921.1 | XP_016871410.1 | - | - | - | |
XM_017015922.1 | XP_016871411.1 | - | - | - | |
XM_017015923.1 | XP_016871412.1 | - | - | - | |
XM_017015924.1 | XP_016871413.1 | - | - | - | |
XM_017015925.1 | XP_016871414.1 | - | - | - | |
GenBank | AB030073.1 | BAA89296.1 | - | - | - |
AB084153.1 | BAC45037.1 | - | - | - | |
AK294026.1 | - | - | - | ||
AK308621.1 | - | - | - | ||
BC037338.2 | - | - | - | ||
BC039243.1 | AAH39243.2 | - | - | - | |
BC096749.1 | - | - | - | ||
EF057068.1 | - | - | - | ||
ENST00000358487.0 | - | - | - | ||
M55614.1 | AAA61188.1 | - | - | - | |
M80634.1 | AAA36147.1 | - | - | - | |
M87770.1 | AAA59470.1 | - | - | - | |
M87771.1 | AAA59471.1 | - | - | - | |
M97193.1 | AAA52449.1 | - | - | - | |
S41878.1 | AAB19323.1 | - | - | - | |
X52832.1 | CAA37014.1 | - | - | - | |
Z71929.1 | CAA96492.1 | - | - | - |
Literature Links: |
FGFR2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |