Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DNMT3A | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001320893.1 | NP_001307822.1 | - | - | - |
NM_022552.4 | NP_072046.2 | 1583146 | c.2504C>T | p.T835M | |
6907608 | c.2536C>T | p.Q846* | |||
7412720 | c.2531_2537delAAGACCA | p.K844fs*7 | |||
5944893 | c.2479-1G>T | p.? | |||
6891590 | c.2490G>A | p.V830V | |||
1318947 | c.2486A>G | p.K829R | |||
6906593 | c.2560G>C | p.E854Q | |||
5944203 | c.2554delA | p.M852fs*1 | |||
1583148 | c.2486A>T | p.K829I | |||
4383531 | c.2524C>G | p.Q842E | |||
4383527 | c.2569G>A | p.D857N | |||
5878746 | c.2544delT | p.P849fs*4 | |||
3743730 | c.2581T>C | p.C861R | |||
5944125 | c.2510_2511insT | p.N838fs*17 | |||
5944887 | c.2589A>C | p.E863D | |||
7107574 | c.2481C>T | p.F827F | |||
6854565 | c.2525A>G | p.Q842R | |||
3719753 | c.2597+1G>A | p.? | |||
7308437 | c.2514_2515insA | p.S839fs*16 | |||
7342027 | c.2566G>T | p.E856* | |||
1169638 | c.2501C>T | p.T834I | |||
7340254 | c.2562G>C | p.E854D | |||
5752684 | c.2524C>T | p.Q842* | |||
1583127 | c.2587G>A | p.E863K | |||
133130 | c.2546C>G | p.P849R | |||
1583125 | c.2553_2553delC | p.F851fs*2 | |||
231551 | c.2580_2597del18 | p.C861_R866del | |||
1583147 | c.2526G>T | p.Q842H | |||
5944895 | c.2479-2A>G | p.? | |||
5427441 | c.2516C>A | p.S839Y | |||
7335400 | c.2568G>C | p.E856D | |||
1169637 | c.2502_2531del30 | p.T835_K844del | |||
1614666 | c.2530A>G | p.K844E | |||
53099 | c.2597+2T>G | p.? | |||
5878748 | c.2479-2A>C | p.? | |||
4383529 | c.2546C>T | p.P849L | |||
221581 | c.2506A>T | p.R836W | |||
5878744 | c.2577A>C | p.L859F | |||
4383525 | c.2576delT | p.L859fs*22 | |||
4766067 | c.2521A>G | p.K841E | |||
5944889 | c.2554A>G | p.M852V | |||
7106437 | c.2501C>A | p.T834N | |||
4383533 | c.2483G>A | p.S828N | |||
231568 | c.2578T>C | p.W860R | |||
1737880 | c.2506_2506delA | p.R836fs*5 | |||
6907681 | c.2576T>G | p.L859* | |||
53096 | c.2481_2481delC | p.F827fs*4 | |||
221578 | c.2485A>T | p.K829* | |||
133731 | c.2510C>G | p.S837* | |||
4169946 | c.2580G>A | p.W860* | |||
6907436 | c.2560G>T | p.E854* | |||
231575 | c.2512A>G | p.N838D | |||
1683137 | c.2571delC | p.D857fs*24 | |||
6907636 | c.2523_2524delGC | p.Q842fs*12 | |||
1583126 | c.2551_2552delTT | p.F851fs*3 | |||
6954732 | c.2592G>A | p.M864I | |||
NM_153759.3 | NP_715640.2 | 6891591 | c.1923G>A | p.V641V | |
1614667 | c.1963A>G | p.K655E | |||
3743731 | c.2014T>C | p.C672R | |||
1318948 | c.1919A>G | p.K640R | |||
7340255 | c.1995G>C | p.E665D | |||
5150506 | c.1912-3T>C | p.? | |||
5351697 | c.1945A>G | p.N649D | |||
5878747 | c.1977delT | p.P660fs*4 | |||
4383530 | c.1979C>T | p.P660L | |||
3991221 | c.1937C>T | p.T646M | |||
7106438 | c.1934C>A | p.T645N | |||
4383526 | c.2009delT | p.L670fs*22 | |||
1683138 | c.2004delC | p.D668fs*24 | |||
7132986 | c.2020G>A | p.E674K | |||
7339850 | c.1943C>G | p.S648* | |||
4383534 | c.1916G>A | p.S639N | |||
6907609 | c.1969C>T | p.Q657* | |||
7308438 | c.1947_1948insA | p.S650fs*16 | |||
6954733 | c.2025G>A | p.M675I | |||
4169947 | c.2013G>A | p.W671* | |||
6945287 | c.1959G>T | p.Q653H | |||
5752685 | c.1957C>T | p.Q653* | |||
5427442 | c.1949C>A | p.S650Y | |||
6907682 | c.2009T>G | p.L670* | |||
5878745 | c.2010A>C | p.L670F | |||
3719754 | c.2030+1G>A | p.? | |||
5878749 | c.1912-2A>C | p.? | |||
7107575 | c.1914C>T | p.F638F | |||
6907637 | c.1956_1957delGC | p.Q653fs*12 | |||
4383528 | c.2002G>A | p.D668N | |||
4383524 | c.2011T>C | p.W671R | |||
4383532 | c.1957C>G | p.Q653E | |||
NM_175629.2 | NP_783328.1 | - | - | - | |
NR_135490.1 | - | - | - | ||
XM_005264175.4 | XP_005264232.1 | - | - | - | |
XM_005264177.4 | XP_005264234.1 | - | - | - | |
XM_011532662.2 | XP_011530964.1 | - | - | - | |
XM_011532663.2 | XP_011530965.1 | - | - | - | |
XM_011532666.2 | XP_011530968.1 | - | - | - | |
XM_011532667.2 | XP_011530969.1 | - | - | - | |
XM_017003526.1 | XP_016859015.1 | - | - | - | |
XM_017003527.1 | XP_016859016.1 | - | - | - | |
GenBank | AB208833.1 | BAD92070.1 | - | - | - |
AF067972.2 | AAD33084.2 | - | - | - | |
AF331856.1 | AAL57039.1 | - | - | - | |
AF480163.1 | AAN40037.1 | - | - | - | |
AK025230.1 | - | - | - | ||
BC018214.1 | AAH18214.1 | - | - | - | |
BC023612.2 | AAH23612.1 | - | - | - | |
BC043617.1 | AAH43617.1 | - | - | - | |
ENST00000321117.0 | - | - | - |
Literature Links: |
DNMT3A PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |