Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CEBPA | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001285829.1 | NP_001272758.1 | - | - | - |
NM_001287424.1 | NP_001274353.1 | - | - | - | |
NM_001287435.1 | NP_001274364.1 | - | - | - | |
NM_004364.4 | NP_004355.2 | 29229 | c.480_481insGTAA | p.K161fs*10 | |
330605 | c.531_532insG | p.L178fs*143 | |||
1374 | c.494delG | p.R165fs*153 | |||
6023053 | c.556C>T | p.P186S | |||
6964248 | c.551C>G | p.P184R | |||
29525 | c.586_587ins15 | p.H195_P196>insHPHPH | |||
27471 | c.541delT | p.Y181fs*137 | |||
18628 | c.595_596GC>TT | p.A199L | |||
6959220 | c.582_583insG | p.H195fs*126 | |||
27469 | c.600_1055del456 | p.H200_K352>Q | |||
18450 | c.562_564delCCG | p.P188del | |||
18467 | c.577_578insGCCCGC | p.H193_H193insRPH | |||
29620 | c.501_502insAG | p.D168fs*151 | |||
5065109 | c.578_589del12 | p.H193_P196del | |||
5065107 | c.558_566del9 | p.P187_P189del | |||
5620466 | c.542A>C | p.Y181S | |||
29604 | c.611delC | p.P204fs*114 | |||
6942988 | c.568T>C | p.S190P | |||
5065106 | c.548C>A | p.P183Q | |||
4170207 | c.589_590insACCCGC | p.P196_P197insHP | |||
6933743 | c.539C>G | p.P180R | |||
28624 | c.592_684del93 | p.P198_P228del | |||
6926274 | c.538C>T | p.P180S | |||
18646 | c.539C>A | p.P180H | |||
5064941 | c.625_625delC | p.Q209fs*109 | |||
18644 | c.529_530GG>TT | p.G177F | |||
5064973 | c.499_500insG | p.E167fs*3 | |||
18642 | c.523C>G | p.L175V | |||
27482 | c.477_478insGTCCCCC | p.I160fs*12 | |||
5044355 | c.564_566delGCC | p.P189delP | |||
1379 | c.573C>T | p.H191H | |||
4767838 | c.590C>A | p.P197Q | |||
330604 | c.515_516insCACG | p.Q172fs*150 | |||
6964669 | c.596C>T | p.A199V | |||
4131623 | c.521C>T | p.A174V | |||
18634 | c.593_594insGCCGCCCC | p.A199fs*122 | |||
249786 | c.608_608delC | p.P204fs*114 | |||
27472 | c.542_543insA | p.Y181fs*1 | |||
7448245 | c.590_591insACCCGC | p.A199_H200insPA | |||
29032 | c.540T>C | p.P180P | |||
6945285 | c.600C>G | p.H200Q | |||
5064951 | c.543C>A | p.Y181* | |||
18273 | c.555_593del39 | p.P185_P197del | |||
5620465 | c.545A>C | p.Q182P | |||
42117 | c.541_542insTT | p.Y181fs*138 | |||
6942966 | c.502G>A | p.D168N | |||
29512 | c.498_499insG | p.E167fs*3 | |||
5065105 | c.533_548del16 | p.L178fs*135 | |||
1666668 | c.588_589insCACCCG | p.P196_P197insHP | |||
7001139 | c.547_549delCCG | p.P189delP | |||
5446613 | c.568_569insCGCACC | p.P196_P197insHP | |||
29247 | c.623_624insGGGTC | p.F208fs*112 | |||
5065101 | c.496G>T | p.E166* | |||
28625 | c.570_571insGCACCC | p.S190_H191insAP | |||
20936 | c.539delC | p.P180fs*138 | |||
18647 | c.543C>G | p.Y181* | |||
6926536 | c.621G>C | p.Q207H | |||
29241 | c.513_514insA | p.Q172fs*149 | |||
18645 | c.534C>T | p.L178L | |||
33869 | c.568_569insT | p.S190fs*131 | |||
18643 | c.527C>T | p.A176V | |||
6188550 | c.586_587insCGCACC | p.H195_P196insPH | |||
29589 | c.568_594del27 | p.S190_P198del | |||
27483 | c.232_522del291 | p.L78_A174del | |||
249866 | c.480_480delC | p.K161fs*157 | |||
27481 | c.500_502delAGG | p.E167del | |||
GenBank | BC063874.1 | AAH63874.1 | - | - | - |
ENST00000498907.0 | - | - | - | ||
X87248.1 | CAA60698.1 | - | - | - | |
Y11525.1 | CAA72289.1 | - | - | - |
Literature Links: |
CEBPA PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |