Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
CDK8
Entrez Gene ID
1024
Gene Name:
cyclin dependent kinase 8
Gene Aliases
K35
Gene Chromosome Location
Chr.13: on Build GRCh38
UniGene
Hs.382306
Species:
Human
Primer Chromosome Location
Chr.13: 26385214-26385342 on Build GRCh38
View in UCSC browser
Amplicon Length
129
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00519368_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
CDK8
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_001260.2
NP_001251.1
-
-
-
NM_001318368.1
NP_001305297.1
1366129
c.532C>T
p.R178*
4632056
c.584T>C
p.V195A
946371
c.557C>T
p.P186L
6553855
c.519C>T
p.D173D
459306
c.591C>T
p.F197F
1200578
c.598C>T
p.R200*
6215503
c.560T>C
p.L187S
6944484
c.569T>C
p.L190S
1559411
c.578T>G
p.V193G
5376570
c.574C>T
p.P192S
3936280
c.533G>A
p.R178Q
6922039
c.584T>A
p.V195D
5376569
c.568T>G
p.L190V
1247946
c.613_615delCTT
p.L206delL
3467991
c.556C>T
p.P186S
12811
c.565G>A
p.D189N
1677566
c.594G>T
p.W198C
5761609
c.595T>C
p.Y199H
6283865
c.622G>T
p.A208S
2069801
c.518A>G
p.D173G
6935844
c.613C>T
p.L205F
6936256
c.601G>T
p.A201S
XM_005266216.4
XP_005266273.1
-
-
-
XM_011534864.1
XP_011533166.1
-
-
-
XM_011534865.2
XP_011533167.1
-
-
-
XM_017020317.1
XP_016875806.1
-
-
-
XM_017020318.1
XP_016875807.1
-
-
-
GenBank
AK296084.1
-
-
-
AK296875.1
-
-
-
AY702976.1
AAU29339.1
-
-
-
BC069634.1
AAH69634.1
-
-
-
BC094784.1
-
-
-
BC104492.1
AAI04493.1
-
-
-
BC105950.1
AAI05951.1
-
-
-
BC107601.1
AAI07602.1
-
-
-
ENST00000381527.0
-
-
-
HG518164.1
-
-
-
X85753.1
CAA59754.1
-
-
-
Phenotype
MIM: 603184
Literature Links:
CDK8
PubMed Links
SNP ID
rs373487382
rs765207700
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.13: 26385233-26385233 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.13: 26385294-26385294 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
transcription initiation from RNA polymerase II promoter
protein phosphorylation
positive regulation of transcription from RNA polymerase II promoter
regulation of cell cycle
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
RNA polymerase II carboxy-terminal domain kinase activity
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0