Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EZH2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001203247.1 | NP_001190176.1 | - | - | - |
NM_001203248.1 | NP_001190177.1 | - | - | - | |
NM_001203249.1 | NP_001190178.1 | - | - | - | |
NM_004456.4 | NP_004447.2 | 600057 | c.1689G>T | p.P563P | |
1448987 | c.1741G>A | p.A581T | |||
5945121 | c.1702A>G | p.K568E | |||
3259639 | c.1680C>A | p.N560K | |||
4384272 | c.1708C>T | p.Q570* | |||
6907689 | c.1718C>A | p.T573N | |||
4384276 | c.1673-4G>C | p.? | |||
144175 | c.1742C>T | p.A581V | |||
6211883 | c.1689G>A | p.P563P | |||
7409652 | c.1727G>A | p.C576Y | |||
5872167 | c.1784C>T | p.A595V | |||
6880887 | c.1691G>A | p.G564E | |||
7085432 | c.1687C>G | p.P563A | |||
7350341 | c.1697G>C | p.R566P | |||
3259637 | c.1696C>A | p.R566S | |||
4384270 | c.1770T>G | p.C590W | |||
7000507 | c.1694G>A | p.C565Y | |||
6963491 | c.1781G>A | p.G594E | |||
6495163 | c.1780G>T | p.G594* | |||
52997 | c.1728C>G | p.C576W | |||
133043 | c.1718C>T | p.T573I | |||
5879089 | c.1768_1769insT | p.C590fs*8 | |||
53030 | c.1774_1777delACTT | p.T592fs*82 | |||
6495167 | c.1731G>A | p.P577P | |||
133045 | c.1697G>A | p.R566H | |||
485012 | c.1768T>A | p.C590S | |||
5879093 | c.1720A>G | p.K574E | |||
6986371 | c.1775C>G | p.T592S | |||
6657572 | c.1696C>T | p.R566C | |||
6416527 | c.1705G>A | p.A569T | |||
6955647 | c.1717A>G | p.T573A | |||
1448989 | c.1730C>T | p.P577L | |||
53023 | c.1712G>A | p.C571Y | |||
5351723 | c.1697G>T | p.R566L | |||
5944297 | c.1773_1777delTACTT | p.T592fs*4 | |||
5989730 | c.1748G>A | p.R583Q | |||
4384274 | c.1682G>C | p.R561P | |||
5989988 | c.1711T>C | p.C571R | |||
744996 | c.1698C>T | p.R566R | |||
53004 | c.1691_1691delG | p.G564fs*111 | |||
6971248 | c.1725G>T | p.Q575H | |||
133052 | c.1682G>A | p.R561H | |||
5944126 | c.1690G>A | p.G564R | |||
4384268 | c.1779_1780delTG | p.C593fs*4 | |||
7000505 | c.1712G>C | p.C571S | |||
7000509 | c.1688C>T | p.P563L | |||
6854585 | c.1741G>T | p.A581S | |||
6495165 | c.1749A>G | p.R583R | |||
5879091 | c.1747C>T | p.R583* | |||
6861437 | c.1725G>C | p.Q575H | |||
NM_152998.2 | NP_694543.1 | 5989731 | c.1616G>A | p.R539Q | |
5989989 | c.1579T>C | p.C527R | |||
5351724 | c.1565G>T | p.R522L | |||
6971249 | c.1593G>T | p.Q531H | |||
6495166 | c.1617A>G | p.R539R | |||
6211884 | c.1557G>A | p.P519P | |||
5879092 | c.1615C>T | p.R539* | |||
6657573 | c.1564C>T | p.R522C | |||
7000508 | c.1562G>A | p.C521Y | |||
600058 | c.1557G>T | p.P519P | |||
4384269 | c.1647_1648delTG | p.C549fs*4 | |||
1448990 | c.1598C>T | p.P533L | |||
4384273 | c.1576C>T | p.Q526* | |||
4384277 | c.1541-4G>C | p.? | |||
6495168 | c.1599G>A | p.P533P | |||
6880888 | c.1559G>A | p.G520E | |||
5872168 | c.1652C>T | p.A551V | |||
6907690 | c.1586C>A | p.T529N | |||
6978224 | c.1550G>A | p.R517H | |||
6495164 | c.1648G>T | p.G550* | |||
7085433 | c.1555C>G | p.P519A | |||
6861438 | c.1593G>C | p.Q531H | |||
6955648 | c.1585A>G | p.T529A | |||
5879090 | c.1636_1637insT | p.C546fs*8 | |||
485013 | c.1636T>A | p.C546S | |||
5879094 | c.1588A>G | p.K530E | |||
744997 | c.1566C>T | p.R522R | |||
7000506 | c.1580G>C | p.C527S | |||
6963492 | c.1649G>A | p.G550E | |||
6416528 | c.1573G>A | p.A525T | |||
7000510 | c.1556C>T | p.P519L | |||
1448988 | c.1609G>A | p.A537T | |||
4384271 | c.1638T>G | p.C546W | |||
6986372 | c.1643C>G | p.T548S | |||
3259640 | c.1548C>A | p.N516K | |||
4384275 | c.1550G>C | p.R517P | |||
XM_005249962.4 | XP_005250019.1 | - | - | - | |
XM_005249963.4 | XP_005250020.1 | - | - | - | |
XM_005249964.4 | XP_005250021.1 | - | - | - | |
XM_011515883.2 | XP_011514185.1 | - | - | - | |
XM_011515884.2 | XP_011514186.1 | - | - | - | |
XM_011515885.2 | XP_011514187.1 | - | - | - | |
XM_011515886.2 | XP_011514188.1 | - | - | - | |
XM_011515887.2 | XP_011514189.1 | - | - | - | |
XM_011515888.2 | XP_011514190.1 | - | - | - | |
XM_011515889.2 | XP_011514191.1 | - | - | - | |
XM_011515890.2 | XP_011514192.1 | - | - | - | |
XM_011515891.2 | XP_011514193.1 | - | - | - | |
XM_011515892.2 | XP_011514194.1 | - | - | - | |
XM_011515893.2 | XP_011514195.1 | - | - | - | |
XM_011515894.2 | XP_011514196.1 | - | - | - | |
XM_011515895.2 | XP_011514197.1 | - | - | - | |
XM_011515896.2 | XP_011514198.1 | - | - | - | |
XM_011515897.2 | XP_011514199.1 | - | - | - | |
XM_011515898.2 | XP_011514200.1 | - | - | - | |
XM_011515901.2 | XP_011514203.1 | - | - | - | |
XM_017011817.1 | XP_016867306.1 | - | - | - | |
XM_017011818.1 | XP_016867307.1 | - | - | - | |
XM_017011819.1 | XP_016867308.1 | - | - | - | |
XM_017011820.1 | XP_016867309.1 | - | - | - | |
XM_017011821.1 | XP_016867310.1 | - | - | - | |
GenBank | AB208895.1 | BAD92132.1 | - | - | - |
AK092676.1 | - | - | - | ||
AK293239.1 | - | - | - | ||
AK302216.1 | - | - | - | ||
AK314291.1 | - | - | - | ||
BC010858.2 | AAH10858.1 | - | - | - | |
BM974565.1 | - | - | - | ||
ENST00000320356.0 | - | - | - | ||
JF327843.1 | - | - | - | ||
U61145.1 | AAC51520.1 | - | - | - | |
X95653.1 | CAA64955.1 | - | - | - |
Literature Links: |
EZH2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Less than 200 bp |