Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CDH1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001317184.1 | NP_001304113.1 | - | - | - |
NM_001317185.1 | NP_001304114.1 | - | - | - | |
NM_001317186.1 | NP_001304115.1 | - | - | - | |
NM_004360.4 | NP_004351.1 | 7347363 | c.676_687+4del16 | p.? | |
20834 | c.612_620delCTTTATTAT | p.F205_I207delFII | |||
3999899 | c.633A>T | p.T211T | |||
6962800 | c.599C>T | p.P200L | |||
3818343 | c.600C>A | p.P200P | |||
6963992 | c.629delA | p.T211fs*4 | |||
972782 | c.671G>A | p.R224H | |||
3421114 | c.628G>T | p.E210* | |||
6927313 | c.616_628>GTT | p.I206fs*6 | |||
4747423 | c.687+1_687+3delGTA | p.? | |||
7345083 | c.682_683insA | p.Y228fs*1 | |||
6974400 | c.683A>G | p.Y228C | |||
7345539 | c.620delT | p.I207fs*8 | |||
6856785 | c.683_684insA | p.Y228fs*1 | |||
1200400 | c.641T>G | p.L214R | |||
5752253 | c.651_652delAG | p.E218fs*4 | |||
4756925 | c.614T>C | p.F205S | |||
1679222 | c.665G>T | p.R222I | |||
5576278 | c.666_677del12 | p.R222_A226>S | |||
19823 | c.602delC | p.P201fs*14 | |||
5833143 | c.612_613insT | p.I206fs*3 | |||
127093 | c.687+2T>G | p.? | |||
2996752 | c.687_687+1delTG | p.? | |||
5055032 | c.641T>C | p.L214P | |||
6916613 | c.634delG | p.G212fs*3 | |||
19825 | c.687+1_687+2delGT | p.? | |||
4766271 | c.662A>G | p.D221G | |||
6961679 | c.639G>A | p.W213* | |||
1324346 | c.662A>T | p.D221V | |||
7345528 | c.594_601delCACACCCC | p.T199fs*7 | |||
6024074 | c.657_658insT | p.L220fs*3 | |||
4062178 | c.664A>G | p.R222G | |||
4747422 | c.602_628del27 | p.P201_E210>Q | |||
6962661 | c.595_596ins22 | p.I206fs*10 | |||
5029220 | c.625_626delAG | p.E210fs*12 | |||
6950478 | c.615_618TATT>A | p.F205_I206>L | |||
972784 | c.681A>G | p.T227T | |||
6954436 | c.641T>A | p.L214Q | |||
5801554 | c.600_601insGT | p.P201fs*15 | |||
3818344 | c.601C>G | p.P201A | |||
6906098 | c.633_634delAG | p.G212fs*10 | |||
4756926 | c.668A>T | p.E223V | |||
6914373 | c.676G>A | p.A226T | |||
6931557 | c.608_609delGT | p.V204fs*4 | |||
2996751 | c.635G>A | p.G212E | |||
6947543 | c.601_616>G | p.P201_I206>V | |||
6856788 | c.602_603insCC | p.V202fs*14 | |||
1379142 | c.670C>T | p.R224C | |||
6958653 | c.660delG | p.D221fs*29 | |||
7343896 | c.622G>T | p.E208* | |||
6934777 | c.681_687+2delATACACTGT | p.? | |||
3888814 | c.601C>T | p.P201S | |||
5833142 | c.598delC | p.P201fs*14 | |||
6909566 | c.628G>C | p.E210Q | |||
6022224 | c.598_599insAC | p.P200fs*16 | |||
5752778 | c.599_600insGT | p.P201fs*15 | |||
5205432 | c.597_598insC | p.V202fs*7 | |||
5576296 | c.663_671delTAGAGAACG | p.R222_R224delRER | |||
XM_011523488.1 | XP_011521790.1 | - | - | - | |
XM_011523489.1 | XP_011521791.1 | - | - | - | |
GenBank | AB025105.1 | BAA88956.1 | - | - | - |
AB025106.1 | BAA88957.1 | - | - | - | |
AK290012.1 | - | - | - | ||
AK297913.1 | - | - | - | ||
AK309703.1 | - | - | - | ||
AK311198.1 | - | - | - | ||
AK312551.1 | - | - | - | ||
BC141838.1 | - | - | - | ||
BC144283.1 | - | - | - | ||
BC146662.1 | - | - | - | ||
ENST00000261769.0 | - | - | - | ||
EU709494.1 | - | - | - | ||
L08599.1 | AAA61259.1 | - | - | - | |
X12790.1 | CAA31279.1 | - | - | - | |
Z13009.1 | CAA78353.1 | - | - | - | |
Z18923.1 | CAA79356.1 | - | - | - |
Literature Links: |
CDH1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |