Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GNAQ | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_002072.4 | NP_002063.2 | 77782 | c.745G>A | p.E249K |
354690 | c.740G>T | p.R247L | |||
4993367 | c.849C>T | p.I283I | |||
7347413 | c.736-6delT | p.? | |||
4993366 | c.857C>T | p.S286F | |||
7347410 | c.736-6_736-5delTT | p.? | |||
5998448 | c.867C>G | p.V289V | |||
7347281 | c.736-7_736-5delTTT | p.? | |||
6447707 | c.832C>A | p.L278I | |||
3218446 | c.803C>T | p.S268L | |||
7347265 | c.736-8_736-7insT | p.? | |||
5438317 | c.736-5delT | p.? | |||
6968302 | c.760C>T | p.L254F | |||
5415480 | c.745G>C | p.E249Q | |||
6342239 | c.740G>A | p.R247Q | |||
6342238 | c.752G>A | p.S251N | |||
1253351 | c.868G>A | p.D290N | |||
28414 | c.796_808del13 | p.N266fs*4 | |||
6342245 | c.836T>C | p.L279P | |||
6918987 | c.816C>A | p.F272L | |||
6918217 | c.739C>T | p.R247* | |||
3908207 | c.826A>G | p.K276E | |||
6944132 | c.854A>G | p.Y285C | |||
5942964 | c.736-4_736-3insTT | p.? | |||
4605880 | c.758C>T | p.A253V | |||
5738627 | c.744G>T | p.M248I | |||
7171880 | c.767G>C | p.R256T | |||
6342251 | c.739C>G | p.R247G | |||
5611525 | c.816_817CT>AA | p.F272_L273>LI | |||
XM_017014628.1 | XP_016870117.1 | - | - | - | |
GenBank | AF011496.1 | AAB64301.1 | - | - | - |
AF329284.1 | AAG61117.1 | - | - | - | |
AF493896.1 | AAM12610.1 | - | - | - | |
AK298579.1 | - | - | - | ||
BC057777.1 | AAH57777.1 | - | - | - | |
BC067850.1 | - | - | - | ||
BC069520.1 | AAH69520.1 | - | - | - | |
BC075096.2 | AAH75096.1 | - | - | - | |
BC075097.2 | AAH75097.1 | - | - | - | |
ENST00000286548.0 | - | - | - | ||
JN184778.1 | - | - | - | ||
L40629.1 | AAA99950.1 | - | - | - | |
L76256.1 | AAB39498.1 | - | - | - | |
U40038.1 | AAC50363.1 | - | - | - | |
U43083.1 | AAB06875.1 | - | - | - |
Literature Links: |
GNAQ PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |