Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
BLNK | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001114094.1 | NP_001107566.1 | - | - | - |
NM_001258440.1 | NP_001245369.1 | - | - | - | |
NM_001258441.1 | NP_001245370.1 | - | - | - | |
NM_001258442.1 | NP_001245371.1 | - | - | - | |
NM_013314.3 | NP_037446.1 | - | - | - | |
NR_047680.1 | - | - | - | ||
NR_047681.1 | - | - | - | ||
NR_047682.1 | - | - | - | ||
NR_047683.1 | - | - | - | ||
XM_011539728.2 | XP_011538030.1 | - | - | - | |
XM_017016159.1 | XP_016871648.1 | - | - | - | |
GenBank | AF068180.1 | AAC39936.1 | - | - | - |
AF068181.1 | AAC39937.1 | - | - | - | |
AK027779.1 | - | - | - | ||
AK225546.1 | - | - | - | ||
AM180327.1 | CAJ55321.1 | - | - | - | |
AM180328.1 | CAJ55322.1 | - | - | - | |
AM180329.1 | CAJ55323.1 | - | - | - | |
AM180330.1 | CAJ55324.1 | - | - | - | |
AM180331.1 | CAJ55325.1 | - | - | - | |
AM180332.1 | CAJ55326.1 | - | - | - | |
AM180333.1 | CAJ55327.1 | - | - | - | |
AM180334.1 | CAJ55328.1 | - | - | - | |
AM180335.1 | CAJ55329.1 | - | - | - | |
AM180336.1 | CAJ55330.1 | - | - | - | |
AM180337.1 | CAJ55331.1 | - | - | - | |
AM180338.1 | CAJ55332.1 | - | - | - | |
AM180339.1 | CAJ55333.1 | - | - | - | |
AM180340.1 | CAJ55334.1 | - | - | - | |
AM180341.1 | CAJ55335.1 | - | - | - | |
AM180342.1 | CAJ55336.1 | - | - | - | |
AM180343.1 | CAJ55337.1 | - | - | - | |
AM180344.1 | CAJ55338.1 | - | - | - | |
AM180345.1 | CAJ55339.1 | - | - | - | |
AM180346.1 | CAJ55340.1 | - | - | - | |
BC018906.2 | AAH18906.1 | - | - | - | |
BC110298.1 | - | - | - | ||
BF975580.1 | - | - | - | ||
ENST00000224337.0 | - | - | - |
Literature Links: |
BLNK PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Less than 200 bp |