Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ETS1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001143820.1 | NP_001137292.1 | 3445160 | c.1307G>A | p.G436D |
1561527 | c.1423G>A | p.A475T | |||
4681821 | c.1331delA | p.N444fs*>42 | |||
4810363 | c.1362C>T | p.Y454Y | |||
1627862 | c.1340A>G | p.H447R | |||
1704806 | c.1367A>T | p.Y456F | |||
4980249 | c.1312C>T | p.R438C | |||
4997511 | c.1408C>T | p.P470S | |||
6265453 | c.1362C>A | p.Y454* | |||
1251531 | c.1431G>A | p.L477L | |||
5519096 | c.1357C>T | p.R453C | |||
3368286 | c.1304G>A | p.R435H | |||
1475248 | c.1358G>A | p.R453H | |||
3445158 | c.1328A>T | p.K443I | |||
4681819 | c.1371C>T | p.R457R | |||
5950159 | c.1320T>G | p.Y440* | |||
4019237 | c.1450G>A | p.D484N | |||
6283789 | c.1352G>A | p.G451E | |||
5367811 | c.1331_1332insA | p.N444fs*18 | |||
1194190 | c.1426A>C | p.M476L | |||
6445626 | c.1409C>A | p.P470H | |||
4398133 | c.1338C>T | p.I446I | |||
5456090 | c.1323C>T | p.Y441Y | |||
5566118 | c.1370G>A | p.R457H | |||
6656687 | c.1363G>A | p.V455M | |||
NM_001162422.1 | NP_001155894.1 | - | - | - | |
NM_005238.3 | NP_005229.1 | 4681820 | c.1239C>T | p.R413R | |
381347 | c.1219G>T | p.G407W | |||
219386 | c.1230C>T | p.Y410Y | |||
1194191 | c.1294A>C | p.M432L | |||
428689 | c.1226G>A | p.R409H | |||
5367812 | c.1199_1200insA | p.N400fs*18 | |||
3445161 | c.1175G>A | p.G392D | |||
6265454 | c.1230C>A | p.Y410* | |||
5519097 | c.1225C>T | p.R409C | |||
1561528 | c.1291G>A | p.A431T | |||
1627863 | c.1208A>G | p.H403R | |||
1704807 | c.1235A>T | p.Y412F | |||
6656688 | c.1231G>A | p.V411M | |||
4681822 | c.1199delA | p.N400fs*>42 | |||
4019238 | c.1318G>A | p.D440N | |||
4980250 | c.1180C>T | p.R394C | |||
4997512 | c.1276C>T | p.P426S | |||
5950160 | c.1188T>G | p.Y396* | |||
280878 | c.1218G>A | p.A406A | |||
3368287 | c.1172G>A | p.R391H | |||
4398134 | c.1206C>T | p.I402I | |||
1251532 | c.1299G>A | p.L433L | |||
3445159 | c.1196A>T | p.K399I | |||
5456091 | c.1191C>T | p.Y397Y | |||
6283790 | c.1220G>A | p.G407E | |||
5566119 | c.1238G>A | p.R413H | |||
6445627 | c.1277C>A | p.P426H | |||
XM_011542649.2 | XP_011540951.1 | - | - | - | |
XM_011542650.1 | XP_011540952.1 | - | - | - | |
XM_011542651.1 | XP_011540953.1 | - | - | - | |
XM_011542652.1 | XP_011540954.1 | - | - | - | |
XM_017017314.1 | XP_016872803.1 | - | - | - | |
XM_017017315.1 | XP_016872804.1 | - | - | - | |
XM_017017316.1 | XP_016872805.1 | - | - | - | |
XM_017017317.1 | XP_016872806.1 | - | - | - | |
GenBank | AK291840.1 | - | - | - | |
AK301405.1 | - | - | - | ||
AY943926.1 | - | - | - | ||
BC017314.2 | AAH17314.1 | - | - | - | |
BT019452.1 | AAV38259.1 | - | - | - | |
BT019453.1 | AAV38260.1 | - | - | - | |
BX640634.1 | CAE45783.1 | - | - | - | |
CR542254.1 | CAG47050.1 | - | - | - | |
ENST00000319397.0 | - | - | - | ||
J04101.1 | AAA52410.1 | - | - | - | |
X14798.1 | CAA32903.1 | - | - | - |
Literature Links: |
ETS1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |