Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
HNF1A | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_000545.6 | NP_000536.5 | 6925573 | c.821A>T | p.E274V |
97133 | c.834_835ins35 | p.H279fs*75 | |||
24930 | c.775G>C | p.V259L | |||
5532358 | c.822A>C | p.E274D | |||
6005771 | c.832C>T | p.R278W | |||
46424 | c.788G>T | p.R263L | |||
6023927 | c.862_864GGG>TGC | p.G288C | |||
6916344 | c.785T>A | p.V262E | |||
4928776 | c.761T>A | p.L254Q | |||
5609082 | c.770A>G | p.N257S | |||
24914 | c.811_818delCGGCGCAA | p.K273fs*41 | |||
5990159 | c.857A>G | p.Y286C | |||
5661820 | c.802T>A | p.F268I | |||
3666559 | c.812_813insCGG | p.R271_R272insG | |||
935974 | c.862delG | p.P291fs*51 | |||
24925 | c.747_764del18 | p.Q250_G255delQAQGLG | |||
24692 | c.787C>T | p.R263C | |||
24929 | c.788G>A | p.R263H | |||
6005774 | c.784G>A | p.V262M | |||
46423 | c.798_799insAAC | p.N266_W267insN | |||
24933 | c.815G>A | p.R272H | |||
4765399 | c.865_866insC | p.G292fs*25 | |||
5591278 | c.812G>A | p.R271Q | |||
4928420 | c.787C>G | p.R263G | |||
5990158 | c.842T>A | p.L281Q | |||
6973928 | c.849G>A | p.M283I | |||
1359417 | c.865delC | p.P291fs*51 | |||
6681148 | c.818_820delAAG | p.E275delE | |||
1476243 | c.863_864insC | p.P289fs*28 | |||
3666557 | c.811_812insCGG | p.R271>PG | |||
6986912 | c.752C>T | p.A251V | |||
5019699 | c.864G>C | p.G288G | |||
5990200 | c.814C>T | p.R272C | |||
7137846 | c.807C>T | p.A269A | |||
1684349 | c.864delG | p.P291fs*51 | |||
6959268 | c.775G>A | p.V259I | |||
5374393 | c.742C>T | p.P248S | |||
6005773 | c.827C>G | p.A276G | |||
394948 | c.857A>T | p.Y286F | |||
21477 | c.817A>G | p.K273E | |||
5988502 | c.838A>C | p.K280Q | |||
1359416 | c.833G>A | p.R278Q | |||
6286609 | c.738G>T | p.V246V | |||
21482 | c.803T>G | p.F268C | |||
24908 | c.829_837delTTCCGGCAC | p.F277_H279delFRH | |||
5611305 | c.800_801GG>AA | p.W267* | |||
6342261 | c.845C>T | p.A282V | |||
7371358 | c.866C>T | p.P289L | |||
3703791 | c.775G>T | p.V259F | |||
46443 | c.770A>C | p.N257T | |||
24920 | c.814C>A | p.R272S | |||
3384093 | c.854C>T | p.T285M | |||
24927 | c.811C>T | p.R271W | |||
4611384 | c.864_865insC | p.G292fs*25 | |||
7239830 | c.746C>G | p.S249* | |||
6005772 | c.780_781GG>AA | p.E261K | |||
21485 | c.749A>C | p.Q250P | |||
1559372 | c.864G>T | p.G288G | |||
6922179 | c.810C>G | p.N270K | |||
24923 | c.779C>T | p.T260M | |||
336015 | c.757G>T | p.G253W | |||
NM_001306179.1 | NP_001293108.1 | - | - | - | |
XM_005253931.3 | XP_005253988.1 | - | - | - | |
GenBank | AK298422.1 | - | - | - | |
BC104908.1 | AAI04909.1 | - | - | - | |
BC104910.1 | AAI04911.1 | - | - | - | |
BC143483.1 | - | - | - | ||
ENST00000257555.0 | - | - | - | ||
HM116541.1 | - | - | - | ||
HM116542.1 | - | - | - | ||
HM116543.1 | - | - | - | ||
HM116544.1 | - | - | - | ||
HM116545.1 | - | - | - | ||
HM116546.1 | - | - | - | ||
HM116547.1 | - | - | - | ||
HM116548.1 | - | - | - | ||
HM116549.1 | - | - | - | ||
HM116550.1 | - | - | - | ||
HM116551.1 | - | - | - | ||
HM116552.1 | - | - | - | ||
HM116557.1 | - | - | - | ||
HM116558.1 | - | - | - | ||
HM116559.1 | - | - | - | ||
HM116560.1 | - | - | - | ||
HM116561.1 | - | - | - | ||
HM116562.1 | - | - | - | ||
HM116563.1 | - | - | - | ||
HM449088.1 | - | - | - | ||
HM449089.1 | - | - | - | ||
M57732.1 | AAA88077.1 | - | - | - | |
X71346.1 | CAB59201.1 | - | - | - | |
X71347.1 | - | - | - |
Literature Links: |
HNF1A PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |