Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EGFR | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_005228.3 | NP_005219.2 | 6191491 | c.311T>A | p.L104Q |
6047686 | c.299C>A | p.P100H | |||
6925694 | c.389C>T | p.T130I | |||
4680390 | c.293G>A | p.R98Q | |||
6196859 | c.380C>T | p.A127V | |||
5041204 | c.385A>G | p.K129E | |||
4893368 | c.347C>T | p.S116F | |||
29339 | c.271A>C | p.I91L | |||
5575629 | c.391G>A | p.G131R | |||
7084541 | c.361G>T | p.V121F | |||
6005740 | c.353C>T | p.A118V | |||
3639718 | c.303G>T | p.L101F | |||
21683 | c.323G>A | p.R108K | |||
4599146 | c.345T>A | p.N115K | |||
5020545 | c.282C>T | p.N94N | |||
1727959 | c.374A>G | p.Y125C | |||
6969229 | c.353_354CC>TT | p.A118V | |||
6023901 | c.344A>C | p.N115T | |||
4892884 | c.298C>T | p.P100S | |||
6915314 | c.332T>C | p.M111T | |||
6196858 | c.341A>G | p.E114G | |||
6960839 | c.380C>A | p.A127E | |||
6307834 | c.350A>G | p.Y117C | |||
6916179 | c.308A>T | p.N103I | |||
1451536 | c.322A>G | p.R108G | |||
6005739 | c.326G>A | p.G109E | |||
174732 | c.340G>A | p.E114K | |||
42977 | c.315G>T | p.Q105H | |||
6178068 | c.271A>G | p.I91V | |||
4600429 | c.316A>T | p.I106F | |||
3639720 | c.348C>T | p.S116S | |||
NM_201282.1 | NP_958439.1 | 6925696 | c.389C>T | p.T130I | |
6047688 | c.299C>A | p.P100H | |||
6178070 | c.271A>G | p.I91V | |||
6960841 | c.380C>A | p.A127E | |||
5575631 | c.391G>A | p.G131R | |||
6939063 | c.322A>G | p.R108G | |||
5835422 | c.340G>A | p.E114K | |||
6969231 | c.353_354CC>TT | p.A118V | |||
6307836 | c.350A>G | p.Y117C | |||
7309721 | c.374A>G | p.Y125C | |||
6652290 | c.293G>A | p.R98Q | |||
7084543 | c.361G>T | p.V121F | |||
6916181 | c.308A>T | p.N103I | |||
6915316 | c.332T>C | p.M111T | |||
5576118 | c.323G>A | p.R108K | |||
6945491 | c.298C>T | p.P100S | |||
NM_201283.1 | NP_958440.1 | - | - | - | |
NM_201284.1 | NP_958441.1 | 3639721 | c.348C>T | p.S116S | |
4893369 | c.347C>T | p.S116F | |||
5575630 | c.391G>A | p.G131R | |||
6925695 | c.389C>T | p.T130I | |||
7084542 | c.361G>T | p.V121F | |||
4600430 | c.316A>T | p.I106F | |||
174733 | c.340G>A | p.E114K | |||
5020546 | c.282C>T | p.N94N | |||
1727960 | c.374A>G | p.Y125C | |||
4680391 | c.293G>A | p.R98Q | |||
5041205 | c.385A>G | p.K129E | |||
6916180 | c.308A>T | p.N103I | |||
1559805 | c.315G>T | p.Q105H | |||
3639719 | c.303G>T | p.L101F | |||
4892885 | c.298C>T | p.P100S | |||
6047687 | c.299C>A | p.P100H | |||
6960840 | c.380C>A | p.A127E | |||
6178069 | c.271A>G | p.I91V | |||
4599147 | c.345T>A | p.N115K | |||
1451537 | c.322A>G | p.R108G | |||
6969230 | c.353_354CC>TT | p.A118V | |||
6307835 | c.350A>G | p.Y117C | |||
1559806 | c.323G>A | p.R108K | |||
6915315 | c.332T>C | p.M111T | |||
GenBank | AB209442.1 | BAD92679.1 | - | - | - |
AF125253.1 | AAG43240.1 | - | - | - | |
AK127817.1 | - | - | - | ||
AK225422.1 | - | - | - | ||
AK290352.1 | - | - | - | ||
AK294750.1 | - | - | - | ||
AW163038.1 | - | - | - | ||
AY550966.1 | AAT52212.1 | - | - | - | |
AY698023.1 | AAT97978.1 | - | - | - | |
AY698024.1 | AAT97979.1 | - | - | - | |
BC070081.1 | - | - | - | ||
BC094761.1 | AAH94761.1 | - | - | - | |
DQ088980.1 | - | - | - | ||
ENST00000275493.0 | - | - | - | ||
GU255993.1 | - | - | - | ||
HQ912715.1 | - | - | - | ||
K03193.1 | AAA52371.1 | - | - | - | |
U48722.1 | AAC50802.1 | - | - | - | |
U95089.1 | AAB53063.1 | - | - | - | |
X00588.1 | CAA25240.1 | - | - | - |
Literature Links: |
EGFR PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |