Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAMK2D | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001221.3 | NP_001212.2 | 3599306 | c.1299G>A | p.R433R |
3696398 | c.1306C>T | p.Q436* | |||
5459082 | c.1279G>A | p.A427T | |||
3748826 | c.1247A>C | p.N416T | |||
1328723 | c.1329G>A | p.M443I | |||
1578845 | c.1210T>C | p.S404P | |||
1618301 | c.1291T>G | p.Y431D | |||
5817728 | c.1293T>C | p.Y431Y | |||
4121501 | c.1206T>G | p.A402A | |||
732898 | c.1302C>T | p.L434L | |||
6007078 | c.1289C>A | p.A430E | |||
7400427 | c.1332A>G | p.P444P | |||
3365468 | c.1325G>T | p.G442V | |||
6237043 | c.1231C>A | p.H411N | |||
NM_001321566.1 | NP_001308495.1 | - | - | - | |
NM_001321567.1 | NP_001308496.1 | - | - | - | |
NM_001321568.1 | NP_001308497.1 | - | - | - | |
NM_001321569.1 | NP_001308498.1 | - | - | - | |
NM_001321570.1 | NP_001308499.1 | - | - | - | |
NM_001321571.1 | NP_001308500.1 | - | - | - | |
NM_001321572.1 | NP_001308501.1 | - | - | - | |
NM_001321573.1 | NP_001308502.1 | - | - | - | |
NM_001321574.1 | NP_001308503.1 | - | - | - | |
NM_001321575.1 | NP_001308504.1 | - | - | - | |
NM_001321576.1 | NP_001308505.1 | - | - | - | |
NM_001321577.1 | NP_001308506.1 | - | - | - | |
NM_001321578.1 | NP_001308507.1 | - | - | - | |
NM_001321579.1 | NP_001308508.1 | - | - | - | |
NM_001321580.1 | NP_001308509.1 | - | - | - | |
NM_001321581.1 | NP_001308510.1 | - | - | - | |
NM_001321582.1 | NP_001308511.1 | - | - | - | |
NM_001321583.1 | NP_001308512.1 | - | - | - | |
NM_001321584.1 | NP_001308513.1 | - | - | - | |
NM_001321585.1 | NP_001308514.1 | - | - | - | |
NM_001321586.1 | NP_001308515.1 | - | - | - | |
NM_001321587.1 | NP_001308516.1 | - | - | - | |
NM_001321588.1 | NP_001308517.1 | - | - | - | |
NM_001321589.1 | NP_001308518.1 | 3660850 | c.1333T>G | p.Y445D | |
6007079 | c.1331C>A | p.A444E | |||
7400428 | c.1374A>G | p.P458P | |||
3696399 | c.1348C>T | p.Q450* | |||
3748827 | c.1289A>C | p.N430T | |||
4859885 | c.1344C>T | p.L448L | |||
6237044 | c.1273C>A | p.H425N | |||
4121502 | c.1248T>G | p.A416A | |||
5459083 | c.1321G>A | p.A441T | |||
3365469 | c.1367G>T | p.G456V | |||
3599307 | c.1341G>A | p.R447R | |||
5817729 | c.1335T>C | p.Y445Y | |||
NM_001321590.1 | NP_001308519.1 | - | - | - | |
NM_001321591.1 | NP_001308520.1 | 5459084 | c.1312G>A | p.A438T | |
4121503 | c.1239T>G | p.A413A | |||
1578846 | c.1243T>C | p.S415P | |||
1618302 | c.1324T>G | p.Y442D | |||
5817730 | c.1326T>C | p.Y442Y | |||
6007080 | c.1322C>A | p.A441E | |||
3365470 | c.1358G>T | p.G453V | |||
7400429 | c.1365A>G | p.P455P | |||
3599308 | c.1332G>A | p.R444R | |||
6237045 | c.1264C>A | p.H422N | |||
4859886 | c.1335C>T | p.L445L | |||
3696400 | c.1339C>T | p.Q447* | |||
3748828 | c.1280A>C | p.N427T | |||
NM_172114.1 | NP_742112.1 | - | - | - | |
NM_172115.2 | NP_742113.1 | - | - | - | |
NM_172127.2 | NP_742125.1 | - | - | - | |
NM_172128.2 | NP_742126.1 | - | - | - | |
NM_172129.1 | NP_742127.1 | - | - | - | |
XM_005263253.3 | XP_005263310.1 | - | - | - | |
XM_005263254.3 | XP_005263311.1 | - | - | - | |
XM_006714330.2 | XP_006714393.1 | - | - | - | |
XM_011532289.1 | XP_011530591.1 | - | - | - | |
XM_011532290.1 | XP_011530592.1 | - | - | - | |
XM_011532291.1 | XP_011530593.1 | - | - | - | |
XM_011532292.2 | XP_011530594.1 | - | - | - | |
XM_011532295.1 | XP_011530597.1 | - | - | - | |
XM_017008672.1 | XP_016864161.1 | - | - | - | |
XM_017008673.1 | XP_016864162.1 | - | - | - | |
XM_017008674.1 | XP_016864163.1 | - | - | - | |
XM_017008675.1 | XP_016864164.1 | - | - | - | |
XM_017008676.1 | XP_016864165.1 | - | - | - | |
XM_017008677.1 | XP_016864166.1 | - | - | - | |
XM_017008678.1 | XP_016864167.1 | - | - | - | |
GenBank | AB209288.1 | BAD92525.1 | - | - | - |
AF071569.1 | AAD20442.1 | - | - | - | |
AK025368.1 | - | - | - | ||
AK055642.1 | - | - | - | ||
AK294517.1 | - | - | - | ||
AL120285.1 | - | - | - | ||
AY987011.1 | AAX88806.1 | - | - | - | |
BC032784.1 | AAH32784.1 | - | - | - | |
ENST00000342666.0 | - | - | - | ||
U50361.1 | AAB16866.1 | - | - | - | |
U73738.1 | AAB18327.1 | - | - | - |
Literature Links: |
CAMK2D PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |