Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GRB10 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001001549.2 | NP_001001549.1 | - | - | - |
NM_001001550.2 | NP_001001550.1 | - | - | - | |
NM_001001555.2 | NP_001001555.1 | - | - | - | |
NM_005311.4 | NP_005302.3 | - | - | - | |
XM_011515304.1 | XP_011513606.1 | - | - | - | |
XM_011515312.2 | XP_011513614.1 | - | - | - | |
XM_011515316.1 | XP_011513618.1 | - | - | - | |
XM_011515323.2 | XP_011513625.1 | - | - | - | |
XM_017012029.1 | XP_016867518.1 | - | - | - | |
XM_017012030.1 | XP_016867519.1 | - | - | - | |
XM_017012031.1 | XP_016867520.1 | - | - | - | |
XM_017012032.1 | XP_016867521.1 | - | - | - | |
XM_017012033.1 | XP_016867522.1 | - | - | - | |
XM_017012034.1 | XP_016867523.1 | 6050021 | c.1457-10C>A | p.? | |
3639534 | c.1508C>T | p.S503F | |||
3639532 | c.1536C>T | p.L512L | |||
3265592 | c.1521T>C | p.I507I | |||
5436861 | c.1537G>A | p.V513M | |||
3698462 | c.1485C>T | p.H495H | |||
7370161 | c.1525C>G | p.Q509E | |||
4999235 | c.1540G>C | p.D514H | |||
6178130 | c.1504_1505GA>AT | p.E502I | |||
746565 | c.1512C>A | p.H504Q | |||
1090621 | c.1466G>T | p.R489M | |||
6515001 | c.1499G>T | p.R500M | |||
XM_017012035.1 | XP_016867524.1 | - | - | - | |
XM_017012036.1 | XP_016867525.1 | - | - | - | |
XM_017012037.1 | XP_016867526.1 | - | - | - | |
XM_017012038.1 | XP_016867527.1 | - | - | - | |
XM_017012039.1 | XP_016867528.1 | - | - | - | |
XM_017012040.1 | XP_016867529.1 | - | - | - | |
XM_017012041.1 | XP_016867530.1 | - | - | - | |
XM_017012042.1 | XP_016867531.1 | - | - | - | |
XM_017012043.1 | XP_016867532.1 | - | - | - | |
XM_017012044.1 | XP_016867533.1 | - | - | - | |
XM_017012045.1 | XP_016867534.1 | - | - | - | |
XM_017012046.1 | XP_016867535.1 | - | - | - | |
XM_017012047.1 | XP_016867536.1 | - | - | - | |
XM_017012048.1 | XP_016867537.1 | - | - | - | |
XM_017012049.1 | XP_016867538.1 | - | - | - | |
XM_017012050.1 | XP_016867539.1 | - | - | - | |
XM_017012051.1 | XP_016867540.1 | - | - | - | |
XM_017012052.1 | XP_016867541.1 | - | - | - | |
XM_017012053.1 | XP_016867542.1 | - | - | - | |
XM_017012054.1 | XP_016867543.1 | - | - | - | |
XM_017012055.1 | XP_016867544.1 | - | - | - | |
XM_017012056.1 | XP_016867545.1 | - | - | - | |
XM_017012057.1 | XP_016867546.1 | - | - | - | |
XM_017012058.1 | XP_016867547.1 | - | - | - | |
XM_017012059.1 | XP_016867548.1 | - | - | - | |
XM_017012060.1 | XP_016867549.1 | 3639535 | c.1490C>T | p.S497F | |
1090622 | c.1448G>T | p.R483M | |||
3639533 | c.1518C>T | p.L506L | |||
4999236 | c.1522G>C | p.D508H | |||
746566 | c.1494C>A | p.H498Q | |||
6178131 | c.1486_1487GA>AT | p.E496I | |||
3265593 | c.1503T>C | p.I501I | |||
5436862 | c.1519G>A | p.V507M | |||
7370162 | c.1507C>G | p.Q503E | |||
6515002 | c.1481G>T | p.R494M | |||
3698463 | c.1467C>T | p.H489H | |||
6050022 | c.1439-10C>A | p.? | |||
XM_017012061.1 | XP_016867550.1 | - | - | - | |
XM_017012062.1 | XP_016867551.1 | - | - | - | |
XM_017012063.1 | XP_016867552.1 | - | - | - | |
XM_017012064.1 | XP_016867553.1 | - | - | - | |
XM_017012065.1 | XP_016867554.1 | - | - | - | |
XM_017012066.1 | XP_016867555.1 | - | - | - | |
XM_017012067.1 | XP_016867556.1 | - | - | - | |
XM_017012068.1 | XP_016867557.1 | - | - | - | |
GenBank | AB000731.1 | - | - | - | |
AF000017.1 | AAC19748.1 | - | - | - | |
AF000018.1 | AAC19749.1 | - | - | - | |
AF001534.1 | AAB81134.1 | - | - | - | |
AJ271366.1 | CAB96542.1 | - | - | - | |
AK289511.1 | - | - | - | ||
BC024285.1 | AAH24285.1 | - | - | - | |
BX649028.1 | - | - | - | ||
D86962.1 | BAA13198.2 | - | - | - | |
ENST00000398812.0 | - | - | - | ||
U34355.1 | AAA88819.1 | - | - | - | |
U66065.1 | AAC50671.1 | - | - | - | |
U69276.1 | AAB08431.1 | - | - | - |
Literature Links: |
GRB10 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Between 200-400 bp |