Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CELF1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001025596.2 | NP_001020767.1 | 7334278 | c.1354_1380del27 | p.P452_Q460delPVSAQAAIQ |
3447834 | c.1382C>T | p.S461F | |||
927443 | c.1436C>T | p.S479L | |||
927444 | c.1389C>T | p.N463N | |||
1981730 | c.1411C>T | p.R471W | |||
4033195 | c.1390G>A | p.G464S | |||
1628000 | c.1346_1347insAA | p.Y449fs*1 | |||
6626102 | c.1404C>T | p.G468G | |||
5566158 | c.1412G>A | p.R471Q | |||
6626105 | c.1432C>T | p.R478C | |||
3791528 | c.1440G>A | p.K480K | |||
6626121 | c.1347C>T | p.Y449Y | |||
5999487 | c.1372G>T | p.A458S | |||
4876869 | c.1361C>T | p.S454L | |||
6068677 | c.1454C>T | p.P485L | |||
NM_001172639.1 | NP_001166110.1 | 6626120 | c.1425C>T | p.Y475Y | |
6068676 | c.1532C>T | p.P511L | |||
7123154 | c.1518G>A | p.K506K | |||
5566157 | c.1490G>A | p.R497Q | |||
5568355 | c.1439C>T | p.S480L | |||
7334277 | c.1432_1458del27 | p.P478_Q486delPVSAQAAIQ | |||
5612650 | c.1489C>T | p.R497W | |||
6626101 | c.1482C>T | p.G494G | |||
6626104 | c.1510C>T | p.R504C | |||
5999486 | c.1450G>T | p.A484S | |||
NM_001172640.1 | NP_001166111.1 | - | - | - | |
NM_006560.3 | NP_006551.1 | - | - | - | |
NM_198700.2 | NP_941989.1 | 6626122 | c.1338C>T | p.Y446Y | |
5999488 | c.1363G>T | p.A455S | |||
6068678 | c.1445C>T | p.P482L | |||
4865381 | c.1427C>T | p.S476L | |||
3791529 | c.1431G>A | p.K477K | |||
4876870 | c.1352C>T | p.S451L | |||
7334279 | c.1345_1371del27 | p.P449_Q457delPVSAQAAIQ | |||
4033196 | c.1381G>A | p.G461S | |||
4192001 | c.1402C>T | p.R468W | |||
3447835 | c.1373C>T | p.S458F | |||
5566159 | c.1403G>A | p.R468Q | |||
6626103 | c.1395C>T | p.G465G | |||
6626106 | c.1423C>T | p.R475C | |||
XM_011519848.2 | XP_011518150.1 | - | - | - | |
XM_011519850.2 | XP_011518152.2 | - | - | - | |
XM_011519852.2 | XP_011518154.1 | - | - | - | |
XM_011519854.1 | XP_011518156.1 | - | - | - | |
XM_011519855.1 | XP_011518157.1 | - | - | - | |
XM_011519856.1 | XP_011518158.1 | - | - | - | |
XM_011519857.1 | XP_011518159.1 | - | - | - | |
XM_011519858.1 | XP_011518160.1 | - | - | - | |
XM_011519859.1 | XP_011518161.1 | - | - | - | |
XM_017017101.1 | XP_016872590.1 | - | - | - | |
XM_017017102.1 | XP_016872591.1 | - | - | - | |
XM_017017103.1 | XP_016872592.1 | - | - | - | |
XM_017017104.1 | XP_016872593.1 | - | - | - | |
XM_017017105.1 | XP_016872594.1 | - | - | - | |
XM_017017106.1 | XP_016872595.1 | - | - | - | |
XM_017017107.1 | XP_016872596.1 | - | - | - | |
XM_017017108.1 | XP_016872597.1 | - | - | - | |
XM_017017109.1 | XP_016872598.1 | - | - | - | |
XM_017017110.1 | XP_016872599.1 | - | - | - | |
XM_017017111.1 | XP_016872600.1 | - | - | - | |
XM_017017112.1 | XP_016872601.1 | - | - | - | |
XM_017017113.1 | XP_016872602.1 | - | - | - | |
XM_017017114.1 | XP_016872603.1 | - | - | - | |
XM_017017115.1 | XP_016872604.1 | - | - | - | |
XM_017017116.1 | XP_016872605.1 | - | - | - | |
XM_017017117.1 | XP_016872606.1 | - | - | - | |
XM_017017118.1 | XP_016872607.1 | - | - | - | |
XM_017017119.1 | XP_016872608.1 | - | - | - | |
XM_017017120.1 | XP_016872609.1 | - | - | - | |
XM_017017121.1 | XP_016872610.1 | - | - | - | |
XM_017017122.1 | XP_016872611.1 | - | - | - | |
XM_017017123.1 | XP_016872612.1 | - | - | - | |
XM_017017124.1 | XP_016872613.1 | - | - | - | |
XM_017017125.1 | XP_016872614.1 | - | - | - | |
XM_017017126.1 | XP_016872615.1 | - | - | - | |
XM_017017127.1 | XP_016872616.1 | - | - | - | |
XM_017017128.1 | XP_016872617.1 | - | - | - | |
XM_017017129.1 | XP_016872618.1 | - | - | - | |
XM_017017130.1 | XP_016872619.1 | - | - | - | |
XM_017017131.1 | XP_016872620.1 | - | - | - | |
XM_017017132.1 | XP_016872621.1 | - | - | - | |
XM_017017133.1 | XP_016872622.1 | - | - | - | |
XM_017017134.1 | XP_016872623.1 | - | - | - | |
XM_017017135.1 | XP_016872624.1 | - | - | - | |
GenBank | AB210019.1 | - | - | - | |
AF248648.1 | AAF86230.1 | - | - | - | |
AF267533.1 | AAF78955.1 | - | - | - | |
AF267534.1 | AAF78956.1 | - | - | - | |
AJ007988.1 | CAC20566.1 | - | - | - | |
AK056024.1 | - | - | - | ||
AK304430.1 | - | - | - | ||
AK307679.1 | - | - | - | ||
BC031079.1 | AAH31079.1 | - | - | - | |
BC036782.1 | - | - | - | ||
ENST00000358597.0 | - | - | - | ||
U63289.1 | AAC50895.1 | - | - | - |
Literature Links: |
CELF1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |