Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
BPTF | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_004459.6 | NP_004450.3 | - | - | - |
NM_182641.3 | NP_872579.2 | 6848865 | c.1196T>C | p.L399P | |
6612448 | c.1206G>T | p.L402F | |||
6147514 | c.1276G>A | p.E426K | |||
3378295 | c.1184T>C | p.V395A | |||
304429 | c.1281C>G | p.D427E | |||
5853295 | c.1133G>A | p.R378Q | |||
7246395 | c.1216G>C | p.E406Q | |||
6612434 | c.1244A>C | p.E415A | |||
707550 | c.1090C>T | p.L364L | |||
1680023 | c.1105G>A | p.D369N | |||
4665990 | c.1233A>G | p.V411V | |||
7158868 | c.1231G>A | p.V411I | |||
983279 | c.1061A>T | p.Y354F | |||
983281 | c.1095G>T | p.Q365H | |||
983283 | c.1161A>C | p.I387I | |||
983285 | c.1209T>C | p.L403L | |||
1246196 | c.1264_1266delGAG | p.E423delE | |||
XM_005257150.3 | XP_005257207.1 | - | - | - | |
XM_005257151.3 | XP_005257208.1 | - | - | - | |
XM_005257152.2 | XP_005257209.1 | - | - | - | |
XM_005257153.3 | XP_005257210.1 | - | - | - | |
XM_005257154.3 | XP_005257211.1 | - | - | - | |
XM_005257155.3 | XP_005257212.1 | - | - | - | |
XM_005257156.3 | XP_005257213.1 | - | - | - | |
XM_005257157.3 | XP_005257214.1 | - | - | - | |
XM_005257158.3 | XP_005257215.1 | - | - | - | |
XM_005257159.2 | XP_005257216.1 | - | - | - | |
XM_005257160.2 | XP_005257217.1 | - | - | - | |
XM_005257161.3 | XP_005257218.1 | - | - | - | |
XM_011524520.2 | XP_011522822.1 | - | - | - | |
XM_011524521.2 | XP_011522823.1 | - | - | - | |
XM_011524522.2 | XP_011522824.1 | - | - | - | |
XM_011524523.2 | XP_011522825.1 | - | - | - | |
XM_011524525.2 | XP_011522827.1 | - | - | - | |
XM_011524526.2 | XP_011522828.1 | - | - | - | |
XM_017024353.1 | XP_016879842.1 | - | - | - | |
GenBank | AB032251.1 | BAA89208.1 | - | - | - |
AK293224.1 | - | - | - | ||
AK297281.1 | - | - | - | ||
AY282495.1 | AAP22284.1 | - | - | - | |
BC037422.1 | - | - | - | ||
ENST00000306378.0 | - | - | - | ||
U05237.1 | AAA97522.1 | - | - | - |
Literature Links: |
BPTF PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Between 200-400 bp |