Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FAM49B | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001256763.1 | NP_001243692.1 | - | - | - |
NM_016623.4 | NP_057707.3 | - | - | - | |
NR_046359.1 | - | - | - | ||
NR_046360.1 | - | - | - | ||
NR_046361.1 | - | - | - | ||
XM_005250966.3 | XP_005251023.1 | - | - | - | |
XM_011517108.2 | XP_011515410.1 | - | - | - | |
XM_011517109.2 | XP_011515411.1 | - | - | - | |
XM_011517110.2 | XP_011515412.1 | - | - | - | |
XM_011517111.2 | XP_011515413.1 | - | - | - | |
XM_011517112.2 | XP_011515414.1 | - | - | - | |
XM_011517113.2 | XP_011515415.1 | - | - | - | |
XM_011517114.2 | XP_011515416.1 | - | - | - | |
XM_011517115.2 | XP_011515417.1 | - | - | - | |
XM_011517116.2 | XP_011515418.1 | - | - | - | |
XM_011517117.2 | XP_011515419.1 | - | - | - | |
XM_011517118.2 | XP_011515420.1 | - | - | - | |
XM_011517120.2 | XP_011515422.1 | - | - | - | |
XM_011517121.2 | XP_011515423.1 | - | - | - | |
XM_011517122.2 | XP_011515424.1 | - | - | - | |
XM_011517123.2 | XP_011515425.1 | - | - | - | |
XM_011517124.2 | XP_011515426.1 | - | - | - | |
XM_011517125.2 | XP_011515427.1 | - | - | - | |
XM_017013539.1 | XP_016869028.1 | - | - | - | |
XM_017013540.1 | XP_016869029.1 | - | - | - | |
XM_017013541.1 | XP_016869030.1 | - | - | - | |
XM_017013542.1 | XP_016869031.1 | - | - | - | |
XM_017013543.1 | XP_016869032.1 | - | - | - | |
XM_017013544.1 | XP_016869033.1 | - | - | - | |
XM_017013545.1 | XP_016869034.1 | - | - | - | |
XM_017013546.1 | XP_016869035.1 | - | - | - | |
XM_017013547.1 | XP_016869036.1 | - | - | - | |
XM_017013548.1 | XP_016869037.1 | 454013 | c.429T>G | p.D143E | |
1674174 | c.421C>T | p.R141W | |||
375137 | c.352C>A | p.P118T | |||
6409572 | c.367G>T | p.E123* | |||
5497930 | c.432A>G | p.E144E | |||
1743311 | c.388A>C | p.K130Q | |||
5645064 | c.350_351insT | p.P118fs*13 | |||
1096074 | c.422G>A | p.R141Q | |||
XM_017013549.1 | XP_016869038.1 | - | - | - | |
XM_017013550.1 | XP_016869039.1 | - | - | - | |
XM_017013551.1 | XP_016869040.1 | - | - | - | |
XM_017013552.1 | XP_016869041.1 | - | - | - | |
XM_017013553.1 | XP_016869042.1 | - | - | - | |
XM_017013554.1 | XP_016869043.1 | - | - | - | |
XM_017013555.1 | XP_016869044.1 | - | - | - | |
XM_017013556.1 | XP_016869045.1 | - | - | - | |
XM_017013557.1 | XP_016869046.1 | - | - | - | |
XM_017013558.1 | XP_016869047.1 | - | - | - | |
XM_017013559.1 | XP_016869048.1 | - | - | - | |
XM_017013560.1 | XP_016869049.1 | - | - | - | |
XM_017013561.1 | XP_016869050.1 | - | - | - | |
XM_017013562.1 | XP_016869051.1 | - | - | - | |
GenBank | AF208851.1 | AAF64265.1 | - | - | - |
AK001226.1 | BAA91566.1 | - | - | - | |
AK002059.1 | BAA92062.1 | - | - | - | |
AK130372.1 | - | - | - | ||
AY364259.1 | AAQ76818.1 | - | - | - | |
AY598320.1 | AAT06731.1 | - | - | - | |
BC003599.1 | AAH03599.1 | - | - | - | |
BC016345.1 | AAH16345.1 | - | - | - | |
BC017297.2 | AAH17297.1 | - | - | - | |
BX419336.2 | - | - | - | ||
CR749628.1 | CAH18422.1 | - | - | - | |
ENST00000522746.0 | - | - | - |
Literature Links: |
FAM49B PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |