Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DIO2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_000793.5 | NP_000784.2 | - | - | - |
NM_001007023.4 | NP_001007024.1 | - | - | - | |
NM_001242502.2 | NP_001229431.1 | - | - | - | |
NM_001242503.2 | NP_001229432.1 | - | - | - | |
NM_001324462.1 | NP_001311391.1 | - | - | - | |
NM_013989.4 | NP_054644.1 | - | - | - | |
GenBank | AB041843.1 | BAB16838.1 | - | - | - |
AB041844.1 | BAB16839.1 | - | - | - | |
AF093774.1 | AAC95470.1 | - | - | - | |
AK292210.1 | - | - | - | ||
AK311603.1 | - | - | - | ||
BC063118.1 | AAH63118.1 | - | - | - | |
BC074882.2 | AAH74882.1 | - | - | - | |
BC136514.1 | - | - | - | ||
CD691767.1 | - | - | - | ||
ENST00000438257.0 | - | - | - | ||
JK546617.1 | - | - | - | ||
U53506.1 | AAC50663.1 | - | - | - |
Literature Links: |
DIO2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |