Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ERG | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001136154.1 | NP_001129626.1 | - | - | - |
NM_001136155.1 | NP_001129627.1 | - | - | - | |
NM_001243428.1 | NP_001230357.1 | - | - | - | |
NM_001243429.1 | NP_001230358.1 | - | - | - | |
NM_001243432.2 | NP_001230361.1 | 2845106 | c.1054G>A | p.D352N | |
6656067 | c.1067G>A | p.R356Q | |||
4876213 | c.948C>T | p.G316G | |||
5707550 | c.1082G>A | p.R361Q | |||
1414063 | c.986C>T | p.S329L | |||
4458653 | c.1094C>T | p.P365L | |||
244304 | c.1083G>T | p.R361R | |||
5565716 | c.1120C>T | p.R374C | |||
4681623 | c.1044G>C | p.M348I | |||
6363198 | c.964C>T | p.Q322* | |||
3664559 | c.959T>A | p.L320H | |||
5965310 | c.1057G>A | p.E353K | |||
6912187 | c.1159A>G | p.K387E | |||
3550667 | c.969C>T | p.F323F | |||
6942825 | c.1126C>T | p.L376F | |||
1616057 | c.1052C>T | p.P351L | |||
4101567 | c.1092A>G | p.K364K | |||
4991420 | c.981C>T | p.L327L | |||
6947987 | c.1130G>C | p.R377P | |||
5906143 | c.1033G>A | p.E345K | |||
419233 | c.1153A>G | p.M385V | |||
5017248 | c.1056C>G | p.D352E | |||
6973946 | c.958C>T | p.L320F | |||
1414061 | c.1129C>T | p.R377C | |||
2845104 | c.1122C>T | p.R374R | |||
6656069 | c.1123G>A | p.A375T | |||
5715003 | c.1066C>T | p.R356W | |||
4753311 | c.1045A>G | p.T349A | |||
2845108 | c.1005C>T | p.S335S | |||
6559125 | c.1125C>A | p.A375A | |||
6954674 | c.1121G>T | p.R374L | |||
6371499 | c.976G>T | p.E326* | |||
1251478 | c.1121G>A | p.R374H | |||
5392885 | c.1001C>T | p.S334F | |||
6913063 | c.1129C>G | p.R377G | |||
4385205 | c.1089C>G | p.S363R | |||
3550666 | c.1075G>A | p.G359R | |||
5986150 | c.1104C>G | p.N368K | |||
4101566 | c.1137C>T | p.Y379Y | |||
5767962 | c.1070G>A | p.R357H | |||
5858620 | c.1093C>T | p.P365S | |||
6976119 | c.1020A>T | p.E340D | |||
6943054 | c.1108G>A | p.D370N | |||
NM_004449.4 | NP_004440.1 | 48371 | c.1058G>T | p.R353L | |
74541 | c.914C>T | p.S305L | |||
48372 | c.877C>A | p.Q293K | |||
6908571 | c.1037A>G | p.D346G | |||
NM_182918.3 | NP_891548.1 | 4897463 | c.948C>T | p.F316F | |
6363199 | c.943C>T | p.Q315* | |||
6954675 | c.1100G>T | p.R367L | |||
4296147 | c.1101C>T | p.R367R | |||
6976120 | c.999A>T | p.E333D | |||
6943055 | c.1087G>A | p.D363N | |||
5906144 | c.1012G>A | p.E338K | |||
4876214 | c.927C>T | p.G309G | |||
5392886 | c.980C>T | p.S327F | |||
4892325 | c.1108C>T | p.R370C | |||
5965311 | c.1036G>A | p.E346K | |||
4296151 | c.984C>T | p.S328S | |||
6912188 | c.1138A>G | p.K380E | |||
4947227 | c.965C>T | p.S322L | |||
6656070 | c.1102G>A | p.A368T | |||
4907241 | c.1071A>G | p.K357K | |||
4812098 | c.1132A>G | p.M378V | |||
6371500 | c.955G>T | p.E319* | |||
4385206 | c.1068C>G | p.S356R | |||
5707551 | c.1061G>A | p.R354Q | |||
6559126 | c.1104C>A | p.A368A | |||
5565717 | c.1099C>T | p.R367C | |||
4753312 | c.1024A>G | p.T342A | |||
6973947 | c.937C>T | p.L313F | |||
6942826 | c.1105C>T | p.L369F | |||
5858621 | c.1072C>T | p.P358S | |||
5986151 | c.1083C>G | p.N361K | |||
6913064 | c.1108C>G | p.R370G | |||
4781113 | c.1031C>T | p.P344L | |||
6947988 | c.1109G>C | p.R370P | |||
4892085 | c.1054G>A | p.G352R | |||
4296149 | c.1033G>A | p.D345N | |||
5767963 | c.1049G>A | p.R350H | |||
5715004 | c.1045C>T | p.R349W | |||
5017249 | c.1035C>G | p.D345E | |||
4681624 | c.1023G>C | p.M341I | |||
4991421 | c.960C>T | p.L320L | |||
4907109 | c.1116C>T | p.Y372Y | |||
4810512 | c.938T>A | p.L313H | |||
4458654 | c.1073C>T | p.P358L | |||
6656068 | c.1046G>A | p.R349Q | |||
XM_011529486.2 | XP_011527788.1 | - | - | - | |
XM_017028287.1 | XP_016883776.1 | - | - | - | |
XM_017028288.1 | XP_016883777.1 | - | - | - | |
XM_017028289.1 | XP_016883778.1 | - | - | - | |
GenBank | AK297807.1 | - | - | - | |
AK300395.1 | - | - | - | ||
AK301277.1 | - | - | - | ||
AK303518.1 | - | - | - | ||
AK309469.1 | - | - | - | ||
AY204741.1 | AAP41719.1 | - | - | - | |
BC040168.1 | AAH40168.1 | - | - | - | |
ENST00000442448.0 | - | - | - | ||
KP100545.1 | - | - | - |
Literature Links: |
ERG PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |