Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
GPRIN2
LOC107984026
Entrez Gene ID
9721
107984026
Gene Name:
G protein regulated inducer of neurite outgrowth 2
arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
Gene Aliases
GRIN2, KIAA0514
-
Gene Chromosome Location
Chr.10: on Build GRCh38
Chr.10: on Build GRCh38
UniGene
Hs.523375
Species:
Human
Primer Chromosome Location
Chr.10: 46550400-46550175 on Build GRCh38
View in UCSC browser
Amplicon Length
226
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00673482_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
GPRIN2
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_014696.3
NP_055511.2
3867241
c.530C>T
p.A177V
5036725
c.439G>A
p.G147S
5034720
c.466C>A
p.L156M
392829
c.440G>T
p.G147V
1958298
c.391C>T
p.R131W
5820803
c.409C>A
p.R137R
918361
c.523G>A
p.D175N
4942327
c.478G>A
p.G160S
4650199
c.471G>T
p.Q157H
4014303
c.405T>G
p.G135G
3807215
c.541G>A
p.E181K
4608315
c.488G>T
p.G163V
3438012
c.556G>A
p.A186T
3438011
c.522G>A
p.R174R
3438010
c.485C>T
p.S162F
1208797
c.364T>C
p.S122P
XM_005277666.3
XP_005277723.1
-
-
-
XM_011540388.2
XP_011538690.1
-
-
-
XM_011540395.2
XP_011538697.1
-
-
-
XM_011540400.2
XP_011538702.1
-
-
-
XM_017016974.1
XP_016872463.1
-
-
-
XM_017016975.1
XP_016872464.1
-
-
-
XM_017016976.1
XP_016872465.1
-
-
-
XM_017016977.1
XP_016872466.1
-
-
-
XM_017016978.1
XP_016872467.1
-
-
-
XM_017016979.1
XP_016872468.1
-
-
-
XM_017016980.1
XP_016872469.1
-
-
-
XM_017016981.1
XP_016872470.1
-
-
-
XM_017016982.1
XP_016872471.1
-
-
-
XM_017016983.1
XP_016872472.1
-
-
-
XM_017016984.1
XP_016872473.1
-
-
-
GenBank
AB011086.1
BAA25440.2
-
-
-
BC011672.2
AAH11672.1
-
-
-
ENST00000374317.0
-
-
-
Phenotype
MIM: 611240
LOC107984026
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
XM_017017029.1
XP_016872518.1
-
-
-
XM_017017030.1
XP_016872519.1
-
-
-
XM_017017031.1
XP_016872520.1
-
-
-
XM_017017032.1
XP_016872521.1
-
-
-
XM_017017033.1
XP_016872522.1
-
-
-
XM_017017034.1
XP_016872523.1
-
-
-
XM_017017038.1
XP_016872527.1
-
-
-
XM_017017039.1
XP_016872528.1
-
-
-
XM_017017040.1
XP_016872529.1
-
-
-
Phenotype
Literature Links:
GPRIN2
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Process(es)
protein binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0