Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATF7IP | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001286514.1 | NP_001273443.1 | - | - | - |
NM_001286515.1 | NP_001273444.1 | 6382209 | c.1828C>A | p.Q610K | |
5006527 | c.1870C>T | p.R624* | |||
6298060 | c.1926+5G>T | p.? | |||
4205631 | c.1912C>T | p.L638F | |||
4205630 | c.1862T>C | p.L621S | |||
3416664 | c.1816T>G | p.L606V | |||
7356804 | c.1843G>A | p.D615N | |||
4166112 | c.1853T>C | p.L618S | |||
7276586 | c.1823C>T | p.A608V | |||
6505254 | c.1820G>A | p.C607Y | |||
4040537 | c.1824G>A | p.A608A | |||
NM_018179.4 | NP_060649.3 | 2002841 | c.1827G>A | p.A609A | |
6505253 | c.1823G>A | p.C608Y | |||
2002844 | c.1915C>T | p.L639F | |||
7276585 | c.1826C>T | p.A609V | |||
2002843 | c.1865T>C | p.L622S | |||
1733305 | c.1858G>A | p.A620T | |||
3416663 | c.1819T>G | p.L607V | |||
937504 | c.1902G>A | p.R634R | |||
937503 | c.1873C>T | p.R625* | |||
6382208 | c.1831C>A | p.Q611K | |||
937502 | c.1835G>A | p.C612Y | |||
1578716 | c.1855T>C | p.L619L | |||
6298059 | c.1929+5G>T | p.? | |||
7356803 | c.1846G>A | p.D616N | |||
4166111 | c.1856T>C | p.L619S | |||
NM_181352.1 | NP_851997.1 | - | - | - | |
XM_005253424.3 | XP_005253481.1 | - | - | - | |
XM_006719108.3 | XP_006719171.1 | - | - | - | |
XM_006719109.3 | XP_006719172.1 | - | - | - | |
XM_011520754.2 | XP_011519056.1 | - | - | - | |
XM_011520755.2 | XP_011519057.1 | - | - | - | |
XM_011520756.2 | XP_011519058.1 | - | - | - | |
XM_017019638.1 | XP_016875127.1 | - | - | - | |
XM_017019639.1 | XP_016875128.1 | - | - | - | |
GenBank | AF425650.1 | AAO91864.1 | - | - | - |
AJ242978.1 | CAB45135.1 | - | - | - | |
AK001550.1 | BAA91751.1 | - | - | - | |
AK025060.1 | - | - | - | ||
AK027575.1 | - | - | - | ||
AK074868.1 | - | - | - | ||
AK299320.1 | - | - | - | ||
AK304184.1 | - | - | - | ||
AY337596.1 | AAQ92978.1 | - | - | - | |
BC053625.1 | - | - | - | ||
BC063855.1 | AAH63855.1 | - | - | - | |
BX648096.1 | - | - | - | ||
ENST00000261168.0 | - | - | - |
Literature Links: |
ATF7IP PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |