Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FBXW7 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001013415.1 | NP_001013433.1 | 4837614 | c.1618C>A | p.R540R |
1149850 | c.1605G>C | p.T535T | |||
6940711 | c.1582G>T | p.V528L | |||
6961408 | c.1571A>G | p.D524G | |||
5640308 | c.1568C>G | p.S523* | |||
7333371 | c.1502-6G>T | p.? | |||
5344636 | c.1577G>A | p.G526E | |||
1594354 | c.1619G>A | p.R540Q | |||
6936450 | c.1513C>T | p.H505Y | |||
4383979 | c.1505C>T | p.P502L | |||
6925216 | c.1554T>A | p.F518L | |||
6958537 | c.1568C>T | p.S523L | |||
6932436 | c.1593G>A | p.W531* | |||
5432180 | c.1502-2A>G | p.? | |||
6939426 | c.1532_1535delGTTT | p.C511fs*8 | |||
6662534 | c.1556T>C | p.V519A | |||
7001785 | c.1608_1609insGAAC | p.F538fs*39 | |||
1427626 | c.1618C>T | p.R540* | |||
7262884 | c.1578A>G | p.G526G | |||
6934765 | c.1599G>C | p.L533F | |||
6099694 | c.1505C>G | p.P502R | |||
3127928 | c.1523C>T | p.A508V | |||
6050828 | c.1535delT | p.L512fs*8 | |||
4684099 | c.1522G>C | p.A508P | |||
4122751 | c.1522G>A | p.A508T | |||
6662519 | c.1605G>A | p.T535T | |||
1149849 | c.1609G>T | p.E537* | |||
1427643 | c.1523C>A | p.A508D | |||
6662500 | c.1526T>C | p.V509A | |||
NM_018315.4 | NP_060785.2 | 4383978 | c.1619C>T | p.P540L | |
6915528 | c.1445G>A | p.R482Q | |||
7262886 | c.1404A>G | p.G468G | |||
6961409 | c.1397A>G | p.D466G | |||
6958536 | c.1682C>T | p.S561L | |||
4122750 | c.1636G>A | p.A546T | |||
7001784 | c.1722_1723insGAAC | p.F576fs*39 | |||
167197 | c.1732C>T | p.R578* | |||
6854475 | c.1349C>T | p.A450V | |||
206684 | c.1733G>A | p.R578Q | |||
1427642 | c.1637C>A | p.A546D | |||
4837613 | c.1732C>A | p.R578R | |||
95598 | c.1707G>A | p.W569* | |||
6925217 | c.1380T>A | p.F460L | |||
5344635 | c.1691G>A | p.G564E | |||
6939427 | c.1358_1361delGTTT | p.C453fs*8 | |||
7262883 | c.1692A>G | p.G564G | |||
6912482 | c.1444C>T | p.R482* | |||
6934766 | c.1425G>C | p.L475F | |||
78945 | c.1712T>A | p.L571* | |||
6936451 | c.1339C>T | p.H447Y | |||
5640307 | c.1682C>G | p.S561* | |||
6662533 | c.1670T>C | p.V557A | |||
6941559 | c.1331C>T | p.P444L | |||
3127927 | c.1637C>T | p.A546V | |||
6958538 | c.1394C>T | p.S465L | |||
5432179 | c.1616-2A>G | p.? | |||
6936449 | c.1627C>T | p.H543Y | |||
6953840 | c.1349C>A | p.A450D | |||
6050827 | c.1649delT | p.L550fs*8 | |||
6099693 | c.1619C>G | p.P540R | |||
7001787 | c.1434_1435insGAAC | p.F480fs*39 | |||
7333373 | c.1328-6G>T | p.? | |||
6939425 | c.1646_1649delGTTT | p.C549fs*8 | |||
4684098 | c.1636G>C | p.A546P | |||
6940710 | c.1696G>T | p.V566L | |||
6662499 | c.1640T>C | p.V547A | |||
6940712 | c.1408G>T | p.V470L | |||
6934764 | c.1713G>C | p.L571F | |||
6662518 | c.1719G>A | p.T573T | |||
732393 | c.1719G>C | p.T573T | |||
6965776 | c.1435G>T | p.E479* | |||
6961407 | c.1685A>G | p.D562G | |||
6925215 | c.1668T>A | p.F556L | |||
732397 | c.1723G>T | p.E575* | |||
7333370 | c.1616-6G>T | p.? | |||
6932437 | c.1419G>A | p.W473* | |||
NM_033632.3 | NP_361014.1 | - | - | - | |
XM_011532083.1 | XP_011530385.1 | - | - | - | |
XM_011532084.1 | XP_011530386.1 | - | - | - | |
XM_011532085.1 | XP_011530387.1 | - | - | - | |
XM_011532086.1 | XP_011530388.1 | - | - | - | |
XM_011532087.1 | XP_011530389.1 | - | - | - | |
XM_011532088.1 | XP_011530390.1 | - | - | - | |
XM_017008362.1 | XP_016863851.1 | - | - | - | |
GenBank | AB209725.1 | BAD92962.1 | - | - | - |
AF383178.1 | AAK60269.1 | - | - | - | |
AF411971.1 | AAL06290.1 | - | - | - | |
AF411972.1 | AAL06291.1 | - | - | - | |
AK001933.1 | BAA91986.1 | - | - | - | |
AK131383.1 | - | - | - | ||
AK223449.1 | BAD97169.1 | - | - | - | |
AK294568.1 | - | - | - | ||
AK299722.1 | - | - | - | ||
AK315142.1 | - | - | - | ||
AY008274.1 | AAG16640.1 | - | - | - | |
AY033553.1 | AAK57547.1 | - | - | - | |
AY049984.1 | AAL07271.1 | - | - | - | |
BC037320.2 | AAH37320.1 | - | - | - | |
BC059373.1 | - | - | - | ||
BC117244.1 | - | - | - | ||
BC117246.1 | - | - | - | ||
BC143944.1 | - | - | - | ||
CR749305.1 | CAH18160.1 | - | - | - | |
ENST00000281708.0 | 5344633 | c.1931G>A | p.G644E | ||
206685 | c.1973G>A | p.R658Q | |||
6934763 | c.1953G>C | p.L651F | |||
6662531 | c.1910T>C | p.V637A | |||
4837611 | c.1972C>A | p.R658R | |||
22933 | c.1947G>A | p.W649* | |||
22967 | c.1972C>T | p.R658* | |||
4684096 | c.1876G>C | p.A626P | |||
6961406 | c.1925A>G | p.D642G | |||
732391 | c.1959G>C | p.T653T | |||
6958535 | c.1922C>T | p.S641L | |||
5432177 | c.1856-2A>G | p.? | |||
6940709 | c.1936G>T | p.V646L | |||
6662516 | c.1959G>A | p.T653T | |||
6939424 | c.1886_1889delGTTT | p.C629fs*8 | |||
6925214 | c.1908T>A | p.F636L | |||
732395 | c.1963G>T | p.E655* | |||
1427640 | c.1877C>A | p.A626D | |||
7346714 | c.1958C>T | p.T653M | |||
29552 | c.1880T>C | p.V627A | |||
78946 | c.1952T>A | p.L651* | |||
7001783 | c.1962_1963insGAAC | p.F656fs*39 | |||
7262882 | c.1932A>G | p.G644G | |||
7349721 | c.1858C>T | p.P620S | |||
3127925 | c.1877C>T | p.A626V | |||
6050825 | c.1889delT | p.L630fs*8 | |||
6936448 | c.1867C>T | p.H623Y | |||
7333369 | c.1856-6G>T | p.? | |||
6099691 | c.1859C>G | p.P620R | |||
29554 | c.1958delC | p.T653fs*8 | |||
4122749 | c.1876G>A | p.A626T | |||
5640305 | c.1922C>G | p.S641* | |||
4383976 | c.1859C>T | p.P620L |
Literature Links: |
FBXW7 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
Set Membership: |
Between 200-400 bp |