Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
CTSE
Entrez Gene ID
1510
Gene Name:
cathepsin E
Gene Aliases
CATE
Gene Chromosome Location
Chr.1: on Build GRCh38
UniGene
Hs.644082
Species:
Human
Primer Chromosome Location
Chr.1: 206012772-206012537 on Build GRCh38
View in UCSC browser
Amplicon Length
236
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00802808_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
CTSE
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_001317331.1
NP_001304260.1
-
-
-
NM_001910.3
NP_001901.1
6637786
c.792G>T
p.Q264H
5379396
c.833C>T
p.A278V
3803642
c.813C>A
p.F271L
902913
c.826T>C
p.C276R
6637793
c.796G>A
p.G266R
82093
c.846A>T
p.T282T
1748115
c.829C>T
p.Q277*
4027497
c.896A>G
p.N299S
3482391
c.848G>A
p.G283E
3750824
c.870T>C
p.P290P
1338263
c.819C>T
p.S273S
NM_148964.2
NP_683865.1
-
-
-
XM_011509244.1
XP_011507546.1
-
-
-
XM_011509245.1
XP_011507547.1
-
-
-
GenBank
AJ250716.1
CAB82849.1
-
-
-
AJ250717.1
CAB82850.1
-
-
-
AK292057.1
-
-
-
AK298069.1
-
-
-
BC042537.1
AAH42537.1
-
-
-
ENST00000358184.0
-
-
-
J05036.1
AAA52130.1
-
-
-
Phenotype
MIM: 116890
Literature Links:
CTSE
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Function(s)
Process(es)
digestion
protein autoprocessing
antigen processing and presentation of exogenous peptide antigen via MHC class II
protein catabolic process
aspartic-type endopeptidase activity
protein homodimerization activity
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0