Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EZH2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001203247.1 | NP_001190176.1 | - | - | - |
NM_001203248.1 | NP_001190177.1 | - | - | - | |
NM_001203249.1 | NP_001190178.1 | - | - | - | |
NM_004456.4 | NP_004447.2 | 1087043 | c.1634A>C | p.Q545P | |
7366858 | c.1571A>T | p.N524I | |||
744994 | c.1667C>G | p.S556* | |||
5990001 | c.1622G>A | p.C541Y | |||
6980986 | c.1594C>T | p.R532W | |||
7412728 | c.1672+2T>C | p.? | |||
6970079 | c.1637A>T | p.N546I | |||
6930313 | c.1669G>C | p.E557Q | |||
5751758 | c.1582_1583insA | p.C528fs*1 | |||
133051 | c.1657C>T | p.Q553* | |||
6907359 | c.1548C>G | p.D516E | |||
3366844 | c.1633C>T | p.Q545* | |||
4168148 | c.1672+5G>T | p.? | |||
3635736 | c.1605T>G | p.C535W | |||
6495171 | c.1642T>G | p.C548G | |||
5731194 | c.1638T>A | p.N546K | |||
1315856 | c.1642_1643insT | p.C548fs*2 | |||
4682157 | c.1608C>A | p.D536E | |||
6918209 | c.1627A>T | p.I543L | |||
3259641 | c.1636A>C | p.N546H | |||
744992 | c.1657C>G | p.Q553E | |||
5766820 | c.1643G>C | p.C548S | |||
7294156 | c.1565T>C | p.V522A | |||
4384278 | c.1582T>C | p.C528R | |||
5005952 | c.1596G>A | p.R532R | |||
144176 | c.1672+2_1673+3insCCT | p.? | |||
1315854 | c.1588C>A | p.H530N | |||
6949859 | c.1649A>C | p.K550T | |||
122927 | c.1613C>T | p.S538L | |||
2270987 | c.1660T>C | p.C554R | |||
5945123 | c.1642T>C | p.C548R | |||
4168150 | c.1672+5G>A | p.? | |||
7410027 | c.1672+2_1672+3insT | p.? | |||
7412596 | c.1643_1643delG | p.C548fs*127 | |||
1195185 | c.1610G>A | p.S537N | |||
6495169 | c.1672+4_1672+7delAGTA | p.? | |||
5731193 | c.1628T>A | p.I543K | |||
1716032 | c.1559A>C | p.N520T | |||
NM_152998.2 | NP_694543.1 | 6495170 | c.1540+4_1540+7delAGTA | p.? | |
3635737 | c.1473T>G | p.C491W | |||
5766821 | c.1511G>C | p.C504S | |||
7294157 | c.1433T>C | p.V478A | |||
5751759 | c.1450_1451insA | p.C484fs*1 | |||
4168151 | c.1540+5G>A | p.? | |||
122928 | c.1481C>T | p.S494L | |||
6930314 | c.1537G>C | p.E513Q | |||
744995 | c.1535C>G | p.S512* | |||
6918210 | c.1495A>T | p.I499L | |||
4682158 | c.1476C>A | p.D492E | |||
1087044 | c.1502A>C | p.Q501P | |||
7366859 | c.1439A>T | p.N480I | |||
6949860 | c.1517A>C | p.K506T | |||
3259642 | c.1504A>C | p.N502H | |||
6495172 | c.1510T>G | p.C504G | |||
5990002 | c.1490G>A | p.C497Y | |||
4168149 | c.1540+5G>T | p.? | |||
1716033 | c.1427A>C | p.N476T | |||
5005953 | c.1464G>A | p.R488R | |||
3366845 | c.1501C>T | p.Q501* | |||
6970080 | c.1505A>T | p.N502I | |||
744993 | c.1525C>G | p.Q509E | |||
1195186 | c.1478G>A | p.S493N | |||
6980987 | c.1462C>T | p.R488W | |||
4384279 | c.1450T>C | p.C484R | |||
XM_005249962.4 | XP_005250019.1 | - | - | - | |
XM_005249963.4 | XP_005250020.1 | - | - | - | |
XM_005249964.4 | XP_005250021.1 | - | - | - | |
XM_011515883.2 | XP_011514185.1 | - | - | - | |
XM_011515884.2 | XP_011514186.1 | - | - | - | |
XM_011515885.2 | XP_011514187.1 | - | - | - | |
XM_011515886.2 | XP_011514188.1 | - | - | - | |
XM_011515887.2 | XP_011514189.1 | - | - | - | |
XM_011515888.2 | XP_011514190.1 | - | - | - | |
XM_011515889.2 | XP_011514191.1 | - | - | - | |
XM_011515890.2 | XP_011514192.1 | - | - | - | |
XM_011515891.2 | XP_011514193.1 | - | - | - | |
XM_011515892.2 | XP_011514194.1 | - | - | - | |
XM_011515893.2 | XP_011514195.1 | - | - | - | |
XM_011515894.2 | XP_011514196.1 | - | - | - | |
XM_011515895.2 | XP_011514197.1 | - | - | - | |
XM_011515896.2 | XP_011514198.1 | - | - | - | |
XM_011515897.2 | XP_011514199.1 | - | - | - | |
XM_011515898.2 | XP_011514200.1 | - | - | - | |
XM_011515901.2 | XP_011514203.1 | - | - | - | |
XM_017011817.1 | XP_016867306.1 | - | - | - | |
XM_017011818.1 | XP_016867307.1 | - | - | - | |
XM_017011819.1 | XP_016867308.1 | - | - | - | |
XM_017011820.1 | XP_016867309.1 | - | - | - | |
XM_017011821.1 | XP_016867310.1 | - | - | - | |
GenBank | AB208895.1 | BAD92132.1 | - | - | - |
AK092676.1 | - | - | - | ||
AK293239.1 | - | - | - | ||
AK302216.1 | - | - | - | ||
AK314291.1 | - | - | - | ||
BC010858.2 | AAH10858.1 | - | - | - | |
ENST00000320356.0 | - | - | - | ||
JF327843.1 | - | - | - | ||
U61145.1 | AAC51520.1 | - | - | - | |
X95653.1 | CAA64955.1 | - | - | - |
Literature Links: |
EZH2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |