Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4634 |
Gene Name: | myosin light chain 3 |
Gene Aliases: |
CMH8, MLC-lV/sb, MLC1SB, MLC1V, VLC1, VLCl |
Location: |
Chr.3:46857867-46863483 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYL3 | NM_000258.2 | 7 | 836 | NP_000249.1 |
AJ709328.1 | 4 | 365 | ||
BC009790.1 | 7 | 801 | AAH09790.1 | |
M24122.1 | 7 | 792 | AAA59895.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508915 | Chr.3:46814349 - 46913473 on Build GRCh38 | Insertion | MYL3 PTH1R PRSS42 |
nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss | CCDC12 SNORD13P3 KIF9 ALS2CL PRSS50 KLHL18 SMARCC1 PRSS45 PTH1R PRSS46 TMIE PRSS43 SETD2 MYL3 KIF9-AS1 ELP6 SCAP CSPG5 NBEAL2 NRADDP PTPN23 PRSS42 |
dgv8308n54 | Chr.3:46811189 - 46894037 on Build GRCh38 | Loss | PRSS43 MYL3 PTH1R PRSS42 |
Set Membership: |
Intragenic Exonic DGV Variation |