Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 462 |
Gene Name: | serpin family C member 1 |
Gene Aliases: |
AT3, AT3D, ATIII, THPH7 |
Location: |
Chr.1:173903804-173917378 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINC1 | NM_000488.3 | NP_000479.1 | ||
XM_005245198.3 | XP_005245255.1 | |||
AF130100.1 | AAG35525.1 | |||
AK312654.1 | ||||
BC022309.1 | AAH22309.1 | |||
D29832.1 | BAA06212.1 | |||
H71210.1 | ||||
KP780080.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758978 | Chr.1:173841504 - 173986805 on Build GRCh38 | Loss | GAS5 SNORD80 SERPINC1 DARS2 SNORD47 SNORD75 SNORD77 SNORD76 SNORA103 RC3H1 SNORD44 SNORD74 GAS5-AS1 SNORD81 SNORD79 ZBTB37 SNORD78 |
nsv831926 | Chr.1:173733264 - 173925529 on Build GRCh38 | Gain+Loss | GAS5 SNORD80 SERPINC1 DARS2 SNORD47 SNORD75 SNORD77 SNORD76 SNORA103 KLHL20 SNORD44 SNORD74 GAS5-AS1 SNORD81 SNORD79 ZBTB37 SNORD78 CENPL |
Set Membership: |
Intragenic Non-exonic DGV Variation |