Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4634 |
Gene Name: | myosin light chain 3 |
Gene Aliases: |
CMH8, MLC-lV/sb, MLC1SB, MLC1V, VLC1, VLCl |
Location: |
Chr.3:46857867-46863483 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYL3 | NM_000258.2 | NP_000249.1 | ||
AF174483.1 | AAF91089.1 | |||
AJ709328.1 | ||||
AK312044.1 | ||||
BC009790.1 | AAH09790.1 | |||
M24122.1 | AAA59895.1 | |||
X07373.1 | CAA30292.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508915 | Chr.3:46814349 - 46913473 on Build GRCh38 | Insertion | PRSS42 PTH1R MYL3 |
nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss | SETD2 SCAP PRSS43 PRSS50 SNORD13P3 TMIE ELP6 ALS2CL MYL3 PRSS45 PTPN23 SMARCC1 KLHL18 PRSS42 CSPG5 NRADDP KIF9 PTH1R KIF9-AS1 CCDC12 NBEAL2 PRSS46 |
dgv8308n54 | Chr.3:46811189 - 46894037 on Build GRCh38 | Loss | PRSS42 PRSS43 PTH1R MYL3 |
Set Membership: |
Intragenic Non-exonic DGV Variation |