Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 9990 |
Gene Name: | solute carrier family 12 member 6 |
Gene Aliases: |
ACCPN, KCC3, KCC3A, KCC3B |
Location: |
Chr.15:34229996-34338064 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC12A6 | NM_001042494.1 | 26 | 4185 | NP_001035959.1 |
NM_001042495.1 | 26 | 3860 | NP_001035960.1 | |
NM_001042496.1 | 26 | 4251 | NP_001035961.1 | |
NM_001042497.1 | 24 | 3954 | NP_001035962.1 | |
NM_005135.2 | 25 | 3824 | NP_005126.1 | |
NM_133647.1 | 25 | 3999 | NP_598408.1 | |
XM_006720793.3 | 24 | 4734 | XP_006720856.1 | |
AF105366.1 | 25 | 3999 | AAD39742.1 | |
AF108831.1 | AAD25337.1 | |||
AF477977.1 | 26 | 4192 | AAL85335.1 | |
AF531258.1 | 24 | 3954 | AAQ10026.1 | |
AF531259.1 | 26 | 4192 | AAQ10027.1 | |
AF531260.1 | 26 | 3830 | AAQ10028.1 | |
AK128133.1 | 24 | 3507 | ||
AL117500.1 | 22 | 3427 | CAB55965.1 | |
BC051744.1 | 26 | 4217 | AAH51744.1 | |
BC070107.1 | 26 | 3855 | ||
BX648195.1 | 25 | 4871 | CAI46042.1 | |
DQ138323.1 | 26 | 4250 | ABA02873.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1052620 | Chr.15:32612402 - 34538731 on Build GRCh38 | Gain | LOC101928042 PGBD4 GOLGA8B LOC105370757 KATNBL1 LOC101928134 MIR1233-2 SLC12A6 NOP10 RYR3 EMC7 NUTM1 FMN1 CHRM5 LPCAT4 GREM1 ARHGAP11A LOC100131315 MIR1233-1 EMC4 TMCO5B AVEN GOLGA8A SCG5 |
esv33337 | Chr.15:31689863 - 34575097 on Build GRCh38 | Gain+Loss | LOC101928042 LOC100996255 PGBD4 LOC105370940 ULK4P1 GOLGA8B LOC105370757 KATNBL1 LOC101928134 MIR1233-2 SLC12A6 NOP10 NUTM1 LPCAT4 GOLGA8K LOC100653133 LOC100131315 MIR1233-1 EMC4 GOLGA8A LOC101060588 WHAMMP1 GOLGA8N RYR3 EMC7 FMN1 CHRM5 GOLGA8O GREM1 ARHGAP11A TMCO5B CHRNA7 AVEN SCG5 |
esv3569431 | Chr.15:33564570 - 35080307 on Build GRCh38 | Gain | AQR PGBD4 GOLGA8B ZNF770 KATNBL1 GJD2 MIR1233-2 SLC12A6 NOP10 RYR3 EMC7 NUTM1 CHRM5 LPCAT4 ACTC1 LOC101928174 MIR1233-1 EMC4 AVEN GOLGA8A |
dgv4456n54 | Chr.15:32604149 - 34557061 on Build GRCh38 | Gain | LOC101928042 PGBD4 GOLGA8B LOC105370757 KATNBL1 LOC101928134 MIR1233-2 SLC12A6 NOP10 NUTM1 LPCAT4 LOC100131315 MIR1233-1 EMC4 GOLGA8A GOLGA8N RYR3 EMC7 FMN1 CHRM5 GREM1 ARHGAP11A TMCO5B AVEN SCG5 |
Set Membership: |
Intragenic Exonic DGV Variation |