Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 10806 |
Gene Name: | serologically defined colon cancer antigen 8 |
Gene Aliases: |
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP |
Location: |
Chr.1:243255328-243500092 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SDCCAG8 | NM_006642.3 | 11 | 1475 | NP_006633.1 |
XM_005273013.4 | 10 | 1307 | XP_005273070.1 | |
XM_005273018.1 | 10 | 1314 | XP_005273075.1 | |
XM_005273021.4 | 11 | 1247 | XP_005273078.1 | |
XM_005273022.3 | 8 | 1096 | XP_005273079.1 | |
XM_006711727.3 | 11 | 1361 | XP_006711790.1 | |
XM_006711728.3 | 10 | 1232 | XP_006711791.1 | |
XM_011544021.2 | 12 | 1457 | XP_011542323.1 | |
XM_011544022.2 | 12 | 1532 | XP_011542324.1 | |
XM_011544023.2 | 12 | 1457 | XP_011542325.1 | |
XM_011544024.2 | 12 | 1457 | XP_011542326.1 | |
XM_011544026.2 | 12 | 1457 | XP_011542328.1 | |
XM_011544028.2 | 10 | 1232 | XP_011542330.1 | |
XM_011544030.2 | 4 | 2008 | XP_011542332.1 | |
XM_017000103.1 | 12 | 1457 | XP_016855592.1 | |
XM_017000104.1 | 10 | 1307 | XP_016855593.1 | |
XM_017000105.1 | 11 | 1462 | XP_016855594.1 | |
XM_017000106.1 | 11 | 1443 | XP_016855595.1 | |
XM_017000107.1 | 12 | 1359 | XP_016855596.1 | |
XM_017000108.1 | 14 | 1906 | XP_016855597.1 | |
AF039690.1 | 5 | 646 | AAC18039.1 | |
AF250731.1 | 11 | 1306 | AAO27830.1 | |
AK297105.1 | 7 | 894 | ||
AK299212.1 | 11 | 1424 | ||
AK299249.1 | 8 | 1067 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833070 | Chr.1:243339809 - 243537501 on Build GRCh38 | Gain | MIR4677 SDCCAG8 AKT3 |
nsv1001488 | Chr.1:243227523 - 243419972 on Build GRCh38 | Loss | MIR4677 SDCCAG8 CEP170 |
nsv517939 | Chr.1:243213455 - 243359604 on Build GRCh38 | Gain | MIR4677 SDCCAG8 CEP170 |
Set Membership: |
Intragenic Exonic DGV Variation |