Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2189 |
Gene Name: | Fanconi anemia complementation group G |
Gene Aliases: |
FAG, XRCC9 |
Location: |
Chr.9:35073838-35080016 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FANCG | NM_004629.1 | 12 | 2074 | NP_004620.1 |
AJ007669.1 | 12 | 2074 | CAA07602.1 | |
AK293427.1 | 12 | 1612 | ||
AK311348.1 | 11 | 1548 | ||
AK312987.1 | 12 | 1999 | ||
BC000032.2 | 12 | 1720 | AAH00032.1 | |
BC011623.2 | 12 | 1847 | AAH11623.1 | |
BT009813.1 | 12 | 1582 | AAP88815.1 | |
KU159422.1 | 12 | 2087 | ||
KU178053.1 | 12 | 1582 | ||
KU178054.1 | 12 | 1564 | ||
KU178055.1 | 12 | 1582 | ||
KU178056.1 | 9 | 1156 | ||
KU178057.1 | ||||
U70310.1 | 12 | 1716 | AAB80802.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |