Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8929 |
Gene Name: | paired like homeobox 2b |
Gene Aliases: |
NBLST2, NBPhox, PMX2B |
Location: |
Chr.4:41744082-41748970 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHOX2B | NM_003924.3 | 1 | 233 | NP_003915.2 |
D82344.1 | 1 | 233 | BAA11555.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |