Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4322 |
Gene Name: | matrix metallopeptidase 13 |
Gene Aliases: |
CLG3, MANDP1, MMP-13 |
Location: |
Chr.11:102942992-102955734 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MMP13 | NM_002427.3 | 2 | 335 | NP_002418.1 |
AK222748.1 | 2 | 335 | BAD96468.1 | |
AK292211.1 | 2 | 335 | ||
AK315341.1 | 2 | 333 | ||
AY256443.1 | 2 | 306 | AAP78939.1 | |
AY256444.1 | 2 | 306 | AAP78940.1 | |
BC067522.1 | 2 | 314 | AAH67522.1 | |
BC067523.1 | 2 | 314 | AAH67523.1 | |
BC074807.2 | 2 | 325 | AAH74807.1 | |
BC074808.2 | 2 | 325 | AAH74808.1 | |
X75308.1 | 2 | 310 | CAA53056.1 | |
X81334.1 | 2 | 333 | CAA57108.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv465 | Chr.11:102926638 - 102971487 on Build GRCh38 | Deletion | MMP13 |
Set Membership: |
Intragenic Exonic DGV Variation |