Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 351 |
Gene Name: | amyloid beta precursor protein |
Gene Aliases: |
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2 |
Location: |
Chr.21:25880550-26171128 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APP | NM_000484.3 | 18 | 3182 | NP_000475.1 |
NM_001136016.3 | 17 | 3106 | NP_001129488.1 | |
NM_001136129.2 | 15 | 2789 | NP_001129601.1 | |
NM_001136130.2 | 17 | 3014 | NP_001129602.1 | |
NM_001136131.2 | 16 | 2844 | NP_001129603.1 | |
NM_001204301.1 | 17 | 3128 | NP_001191230.1 | |
NM_001204302.1 | 16 | 3071 | NP_001191231.1 | |
NM_001204303.1 | 15 | 2903 | NP_001191232.1 | |
NM_201413.2 | 17 | 3125 | NP_958816.1 | |
NM_201414.2 | 16 | 2957 | NP_958817.1 | |
A02759.1 | 16 | 2903 | CAA00268.1 | |
AF282245.1 | 15 | 2716 | AAQ14327.1 | |
AK297412.1 | ||||
BC065529.1 | 17 | 3056 | AAH65529.1 | |
BE907745.1 | ||||
BM876312.1 | 1 | 467 | ||
CB854706.1 | 1 | 466 | ||
M15533.1 | 3 | 963 | AAA35540.1 | |
M18734.1 | 7 | 1465 | AAA51726.1 | |
X06989.1 | 17 | 3049 | CAA30050.1 | |
Y00264.1 | 16 | 2904 | CAA68374.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |