Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 79628 |
Gene Name: | SH3 domain and tetratricopeptide repeats 2 |
Gene Aliases: |
CMT4C, MNMN |
Location: |
Chr.5:148982150-149063174 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SH3TC2 | NM_024577.3 | 17 | 21924 | NP_078853.2 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508383 | Chr.5:148884174 - 148987167 on Build GRCh38 | Deletion | SH3TC2 |
nsv830517 | Chr.5:148953659 - 149171578 on Build GRCh38 | Gain | SH3TC2 ABLIM3 MIR584 LOC255187 |
Set Membership: |
Intragenic Exonic DGV Variation |