Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 26580 |
Gene Name: | BSCL2, seipin lipid droplet biogenesis associated |
Gene Aliases: |
GNG3LG, HMN5, PELD, SPG17 |
Location: |
Chr.11:62690262-62709619 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BSCL2 | NM_001122955.3 | NP_001116427.1 | ||
NR_037948.1 | ||||
NR_037949.1 | ||||
AK315423.1 | ||||
BG699373.1 | ||||
BP370169.1 | ||||
DB178846.1 |
Entrez Gene ID: | 2785 |
Gene Name: | G protein subunit gamma 3 |
Gene Aliases: |
- |
Location: |
Chr.11:62707255-62709206 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNG3 | NM_012202.4 | 1 | 86 | NP_036334.1 |
XM_006718500.2 | XP_006718563.1 | |||
AF087900.1 | AAP97199.1 | |||
AF092129.1 | AAD40371.1 | |||
AK311933.1 | ||||
BC015563.2 | AAH15563.1 | |||
CK004020.1 | ||||
DA123305.1 | 1 | 86 |
Entrez Gene ID: | 100534595 |
Gene Name: | HNRNPUL2-BSCL2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:62690262-62727384 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HNRNPUL2-BSCL2 | NR_037946.1 | |||
AK122942.1 | ||||
CN346952.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv348 | Chr.11:62706978 - 62752240 on Build GRCh38 | Deletion | TTC9C BSCL2 ZBTB3 HNRNPUL2-BSCL2 HNRNPUL2 GNG3 |
nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain | BSCL2 ROM1 EEF1G METTL12 MTA2 HNRNPUL2 EML3 GNG3 UQCC3 B3GAT3 UBXN1 TTC9C C11orf98 HNRNPUL2-BSCL2 GANAB LBHD1 LRRN4CL INTS5 MIR6747 SNORA57 TUT1 |
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |