Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4125 |
Gene Name: | mannosidase alpha class 2B member 1 |
Gene Aliases: |
LAMAN, MANB |
Location: |
Chr.19:12646508-12666777 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAN2B1 | NM_000528.3 | 5 | 763 | NP_000519.2 |
NM_001173498.1 | 5 | 763 | NP_001166969.1 | |
XM_005259913.1 | 5 | 741 | XP_005259970.1 | |
XM_017026818.1 | 3 | 425 | XP_016882307.1 | |
AB209921.1 | 5 | 675 | BAD93158.1 | |
AK291572.1 | 5 | 728 | ||
AK303418.1 | 6 | 702 | ||
AW172353.1 | 4 | 510 | ||
BC000736.2 | 5 | 709 | AAH00736.1 | |
BI828908.1 | ||||
U05572.1 | 5 | 720 | AAB03816.1 | |
U60266.1 | 5 | 701 | AAC34130.1 | |
U68382.1 | 5 | 668 | AAC50811.1 | |
U68567.1 | 5 | 675 | AAC50812.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160586 | Chr.19:12597480 - 12823909 on Build GRCh38 | Duplication | HOOK2 RNASEH2A DHPS ZNF791 JUNB LOC105372280 SNORD41 WDR83OS SNORD135 WDR83 PRDX2 FBXW9 MIR5684 C19orf43 MAN2B1 ZNF490 BEST2 TNPO2 ASNA1 |
dgv138n111 | Chr.19:12627853 - 12810372 on Build GRCh38 | Deletion | HOOK2 RNASEH2A DHPS ZNF791 JUNB LOC105372280 SNORD41 WDR83OS SNORD135 WDR83 PRDX2 FBXW9 MIR5684 C19orf43 MAN2B1 BEST2 TNPO2 ASNA1 |
Set Membership: |
Intragenic Exonic DGV Variation |