Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55737 |
Gene Name: | VPS35 retromer complex component |
Gene Aliases: |
MEM3, PARK17 |
Location: |
Chr.16:46657979-46689232 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VPS35 | NM_018206.5 | 17 | 2508 | NP_060676.2 |
XM_005256045.3 | 16 | 2393 | XP_005256102.1 | |
XM_011523227.2 | 17 | 3238 | XP_011521529.1 | |
AF175265.1 | 17 | 2466 | AAF89953.1 | |
AF183418.1 | 16 | 2417 | AAG09687.1 | |
AF186382.1 | 17 | 2462 | AAG40619.1 | |
AF191298.2 | 17 | 2436 | AAF02778.2 | |
AK000395.1 | 5 | 869 | BAA91137.1 | |
AK001614.1 | 17 | 2456 | BAA91790.1 | |
AK023650.1 | 13 | 2019 | BAB14626.1 | |
AK223218.1 | 17 | 2427 | BAD96938.1 | |
AL136888.1 | 17 | 2456 | CAB66822.1 | |
AL512769.1 | 10 | 1513 | CAC21686.1 | |
AY007112.1 | 6 | 1019 | AAG01989.1 | |
BC002414.2 | 16 | 2415 | AAH02414.1 | |
BC010362.1 | 17 | 2429 | AAH10362.1 | |
BC093036.1 | 17 | 2454 | AAH93036.1 | |
BX647213.1 | 10 | 4245 | CAI46268.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526656 | Chr.16:46505480 - 46812751 on Build GRCh38 | Gain | C16orf87 SHCBP1 MYLK3 VPS35 ANKRD26P1 ORC6 |
nsv428327 | Chr.16:46523056 - 46684578 on Build GRCh38 | Gain | SHCBP1 VPS35 ANKRD26P1 |
dgv2975n100 | Chr.16:46429858 - 46768065 on Build GRCh38 | Gain | SHCBP1 MYLK3 VPS35 ANKRD26P1 ORC6 |
esv3571142 | Chr.16:46430577 - 46840772 on Build GRCh38 | Gain | C16orf87 SHCBP1 MYLK3 VPS35 ANKRD26P1 ORC6 |
esv3638534 | Chr.16:46547097 - 46716009 on Build GRCh38 | Gain | SHCBP1 MYLK3 VPS35 ANKRD26P1 ORC6 |
Set Membership: |
Intragenic Exonic DGV Variation |