Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 1643 |
Gene Name: | damage specific DNA binding protein 2 |
Gene Aliases: |
DDBB, UV-DDB2, XPE |
Location: |
Chr.11:47214942-47239218 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDB2 | NM_000107.2 | 3 | 544 | NP_000098.1 |
NM_001300734.1 | 3 | 544 | NP_001287663.1 | |
AB107037.1 | 3 | 349 | BAD12557.1 | |
AB107038.1 | 3 | 349 | BAD12558.1 | |
AB107039.1 | BAD12559.1 | |||
AB107040.1 | 3 | 349 | BAD12560.1 | |
AK091640.1 | 2 | 1446 | ||
AK313262.1 | 3 | 492 | ||
BC000093.2 | 3 | 372 | AAH00093.1 | |
BT007139.1 | 3 | 349 | AAP35803.1 | |
DB053816.1 | ||||
U18300.1 | 3 | 524 | AAB07897.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |