Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 8292 |
Gene Name: | collagen like tail subunit of asymmetric acetylcholinesterase |
Gene Aliases: |
CMS5, EAD |
Location: |
Chr.3:15450133-15521751 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COLQ | NM_005677.3 | 17 | 2580 | NP_005668.2 |
NM_080538.2 | 17 | 2709 | NP_536799.1 | |
NM_080539.3 | 16 | 2478 | NP_536800.2 | |
AF057036.1 | 17 | 2580 | AAC39927.1 | |
AI246303.1 | ||||
AK128401.1 | 17 | 2709 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834622 | Chr.3:15397807 - 15587083 on Build GRCh38 | Gain | COLQ MIR4270 EAF1 METTL6 HACL1 |
nsv834621 | Chr.3:15324275 - 15509456 on Build GRCh38 | Gain | COLQ SH3BP5 MIR4270 EAF1 METTL6 |
Set Membership: |
Intragenic Exonic DGV Variation |