Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2904 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 2B |
Gene Aliases: |
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3 |
Location: |
Chr.12:13537337-13982012 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRIN2B | NM_000834.3 | 13 | 5284 | NP_000825.2 |
XM_005253351.3 | 4 | 3011 | XP_005253408.1 | |
XM_011520628.2 | 14 | 6077 | XP_011518930.1 | |
XM_011520629.2 | 15 | 5884 | XP_011518931.1 | |
XM_017019219.1 | 14 | 5632 | XP_016874708.1 | |
AB208850.1 | BAD92087.1 | |||
U11287.1 | 13 | 5315 | AAB60368.1 | |
U88963.1 | 13 | 5284 | AAD00659.1 | |
U90278.1 | 13 | 5314 | AAB49993.1 |
Set Membership: |
Intragenic Exonic non-DGV Variation |