Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5631 |
Gene Name: | phosphoribosyl pyrophosphate synthetase 1 |
Gene Aliases: |
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI |
Location: |
Chr.X:107628424-107651026 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRPS1 | NM_001204402.1 | NP_001191331.1 | ||
NM_002764.3 | NP_002755.1 | |||
AK223169.1 | BAD96889.1 | |||
AK297968.1 | ||||
AK307748.1 | ||||
AK312706.1 | ||||
AK316467.1 | ||||
BC001605.1 | AAH01605.1 | |||
D00860.1 | BAA00733.1 | |||
X15331.1 | CAA33386.1 |
Set Membership: |
Intragenic Non-exonic non-DGV Variation |