Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 57654 |
Gene Name: | UV stimulated scaffold protein A |
Gene Aliases: |
KIAA1530, UVSS3 |
Location: |
Chr.4:1345662-1388049 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UVSSA | NM_001317934.1 | NP_001304863.1 | ||
NM_001317935.1 | 1 | 185 | NP_001304864.1 | |
NM_020894.3 | NP_065945.2 | |||
XM_017008490.1 | XP_016863979.1 | |||
XM_017008491.1 | 1 | 125 | XP_016863980.1 | |
XM_017008492.1 | XP_016863981.1 | |||
XM_017008493.1 | XP_016863982.1 | |||
XM_017008494.1 | XP_016863983.1 | |||
XM_017008496.1 | XP_016863985.1 | |||
XM_017008497.1 | XP_016863986.1 | |||
XM_017008499.1 | XP_016863988.1 | |||
AB040963.1 | ||||
BC110331.1 | AAI10332.1 | |||
BC140901.1 | ||||
HY104489.1 | 1 | 185 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv593298 | Chr.4:1250479 - 1348733 on Build GRCh38 | Loss | UVSSA CTBP1-AS2 MAEA |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | MYL5 UVSSA RNF212 LOC100129917 TMEM175 MAEA SLC26A1 FGFRL1 LOC101928548 LOC100130872 LOC107986211 SPON2 LOC105374344 IDUA TMED11P GAK DGKQ CPLX1 CRIPAK CTBP1-AS2 MFSD7 PDE6B LOC101928521 CTBP1-AS CTBP1 ATP5I PCGF3 NKX1-1 |
esv3569247 | Chr.4:1340902 - 1353008 on Build GRCh38 | Loss | UVSSA |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | LOC101928548 UVSSA LOC100130872 CTBP1-AS SPON2 CTBP1 CRIPAK CTBP1-AS2 MAEA NKX1-1 |
nsv593301 | Chr.4:1306289 - 1358897 on Build GRCh38 | Loss | UVSSA MAEA |
nsv593302 | Chr.4:1344830 - 1420896 on Build GRCh38 | Loss | UVSSA CRIPAK NKX1-1 |
nsv508988 | Chr.4:1190955 - 1358870 on Build GRCh38 | Insertion | LOC101928548 UVSSA LOC100130872 CTBP1-AS SPON2 CTBP1 CTBP1-AS2 MAEA |
nsv593281 | Chr.4:1209477 - 1358897 on Build GRCh38 | Loss | UVSSA CTBP1-AS CTBP1 CTBP1-AS2 MAEA |
Set Membership: |
Intragenic Exonic DGV Variation |