Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55063 |
Gene Name: | zinc finger CW-type and PWWP domain containing 1 |
Gene Aliases: |
ZCW1 |
Location: |
Chr.7:100400826-100429239 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZCWPW1 | NM_001258008.1 | NP_001244937.1 | ||
NM_017984.4 | NP_060454.3 | |||
XM_005250479.2 | XP_005250536.1 | |||
XM_005250480.2 | XP_005250537.1 | |||
XM_006716035.3 | XP_006716098.1 | |||
XM_006716036.2 | XP_006716099.1 | |||
XM_006716037.3 | XP_006716100.1 | |||
XM_006716038.3 | XP_006716101.1 | |||
XM_006716040.3 | XP_006716103.1 | |||
XM_017012379.1 | XP_016867868.1 | |||
XM_017012380.1 | XP_016867869.1 | |||
AK000919.1 | BAA91424.1 | |||
AK093038.1 | BAC04028.1 | |||
AK300745.1 | ||||
AK302108.1 | ||||
AL136735.1 | CAB66669.1 | |||
BC002725.1 | AAH02725.1 | |||
KU178677.1 | ||||
KU178678.1 | ||||
KU178679.1 | ||||
KU178680.1 | ||||
KU178681.1 | ||||
KU178682.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | SLC12A9 EPHB4 LAMTOR4 MUC12 AGFG2 GPC2 MUC3A MUC17 AZGP1 GIGYF1 FBXO24 NAT16 PMS2P1 NYAP1 LOC105375429 MIR6875 UFSP1 AP1S1 SRRT LOC102724094 PVRIG2P STAG3L5P MIR106B MOGAT3 MIR6840 ACTL6B EPO SAP25 ACHE PILRB GAL3ST4 MCM7 AP4M1 ZAN SERPINE1 PCOLCE ZKSCAN1 TFR2 MIR93 GATS MBLAC1 TRIP6 ZNHIT1 CLDN15 PILRA LRCH4 GNB2 C7orf43 PLOD3 COPS6 MIR4653 STAG3 PCOLCE-AS1 POP7 ZNF3 SPDYE3 AZGP1P1 CNPY4 PVRIG MIR4658 MOSPD3 C7orf61 PPP1R35 VGF STAG3L5P-PVRIG2P-PILRB TRIM56 MIR25 TSC22D4 ZCWPW1 MEPCE ZSCAN21 TAF6 ZASP FIS1 |
nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss | SPDYE3 PVRIG2P STAG3L5P MIR6840 C7orf61 PPP1R35 STAG3L5P-PVRIG2P-PILRB PILRB PMS2P1 NYAP1 PILRA TSC22D4 ZCWPW1 MEPCE |
Set Membership: |
Intragenic Non-exonic DGV Variation |